EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Hedgehog-regulated atypical PKC promotes phosphorylation and activation of Smoothened and Cubitus interruptus in Drosophila.
PMID 25349414
Abstract
Smoothened (Smo) is essential for transduction of the Hedgehog (Hh) signal in both insects and vertebrates. Cell surface/cilium accumulation of Smo is thought to play an important role in Hh signaling, but how the localization of Smo is controlled remains poorly understood. In this study, we demonstrate that atypical PKC (aPKC) regulates Smo phosphorylation and basolateral accumulation in Drosophila wings. Inactivation of aPKC by either RNAi or a mutation inhibits Smo basolateral accumulation and attenuates Hh target gene expression. In contrast, expression of constitutively active aPKC elevates basolateral accumulation of Smo and promotes Hh signaling. The aPKC-mediated phosphorylation of Smo at Ser680 promotes Ser683 phosphorylation by casein kinase 1 (CK1), and these phosphorylation events elevate Smo activity in vivo. Moreover, aPKC has an additional positive role in Hh signaling by regulating the activity of Cubitus interruptus (Ci) through phosphorylation of the Zn finger DNA-binding domain. Finally, the expression of aPKC is up-regulated by Hh signaling in a Ci-dependent manner. Our findings indicate a direct involvement of aPKC in Hh signaling beyond its role in cell polarity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2104731
Anti-PRKCG antibody produced in rabbit, affinity isolated antibody
SAB2104573
Anti-PRKCG, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2501172 Anti-ptc1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1411779
ANTI-PTCH antibody produced in mouse, clone 1D9, purified immunoglobulin, buffered aqueous solution
SAB1411866
ANTI-PTCH antibody produced in mouse, clone 8A10, purified immunoglobulin, buffered aqueous solution
SAB1411867
ANTI-PTCH antibody produced in mouse, clone 1B7, purified immunoglobulin, buffered aqueous solution
SAB1411868
ANTI-PTCH antibody produced in mouse, clone 3B7, purified immunoglobulin, buffered aqueous solution
SAB1411869
ANTI-PTCH antibody produced in mouse, clone 3C2, purified immunoglobulin, buffered aqueous solution
SAB1411871
ANTI-PTCH antibody produced in mouse, clone 3B3, purified immunoglobulin, buffered aqueous solution
SAB1411872
ANTI-PTCH antibody produced in mouse, clone 4F11, purified immunoglobulin, buffered aqueous solution
AV44249
Anti-PTCH1 antibody produced in rabbit, affinity isolated antibody
SAB1412475
ANTI-SMO antibody produced in mouse, clone 2D10, purified immunoglobulin, buffered aqueous solution
SAB1412476
ANTI-SMO antibody produced in mouse, clone 4E5, purified immunoglobulin, buffered aqueous solution
SAB1412477
ANTI-SMO antibody produced in mouse, clone 4E9, purified immunoglobulin, buffered aqueous solution
WH0005727M2
Monoclonal Anti-PTCH antibody produced in mouse, clone 1G2, purified immunoglobulin, buffered aqueous solution
SAB1404271
Monoclonal Anti-PTCH antibody produced in mouse, clone 8E1, purified immunoglobulin, buffered aqueous solution
WH0006608M1 Monoclonal Anti-SMO antibody produced in mouse, clone 1D6, purified immunoglobulin, buffered aqueous solution