EMAIL THIS PAGE TO A FRIEND
The Nurse practitioner
Drug updates and approvals: 2014 in review.
PMID 25350291
Abstract
This article highlights important prescribing information for new drugs approved by the FDA over the last year. These include albiglutide, apremilast, dapagliflozin, insulin human inhalation powder, riociguat, timothy grass pollen allergen extract, umeclidinium and vilanterol inhalation powder, and vorapaxar.