EMAIL THIS PAGE TO A FRIEND
Biochemistry
BIN1 membrane curvature sensing and generation show autoinhibition regulated by downstream ligands and PI(4,5)P2.
PMID 25350771
Abstract
In striated muscles, invaginations from the plasma membrane, termed transverse tubules (T-tubule), function in the excitation-contraction coupling machinery. BIN1 (isoform8) plays a critical role in the biogenesis of T-tubules. BIN1 contains an N-terminal BAR domain to sense and induce membrane curvature, an isoform8-specific polybasic motif (exon10) as the phosphoinositide binding module and a C-terminal Src homology 3 (SH3) domain for the recruitment of downstream proteins such as dynamin 2. Previous studies of N-BAR domains focused on elucidating mechanisms of membrane curvature sensing and generation (MC-S&G). Less is known about how MC-S&G is regulated. We found that the SH3 domain binds to the exon10 motif more strongly compared to the proline-rich domain (PRD) of dynamin 2. Furthermore, we found that the MC-S&G ability of full-length BIN1 is inhibited on membranes lacking PI(4,5)P2. Addition of PI(4,5)P2 in the membrane activates BIN1 to sense and induce membrane curvature. Co-presence of the SH3 domain and exon10 motif leads to the strongest phosphoinositide-mediated control of BIN1 function. Addition of SH3 domain ligand (such as PRD peptides), as well as addition of the water-soluble PI(4,5)P2 analogue, can both enhance the MC-S&G ability of BIN1 on membranes without PI(4,5)P2, indicating that the key to activate BIN1 is to disrupt the exon10-SH3 interaction. The nonsense mutation K436X, found in centronuclear myopathy (CNM) patients, abolishes SH3 domain binding with either exon10 or the PRD motif, resulting in increased membrane deformation capacity. Our results suggest an autoinhibition model for BIN1 that involves a synergistic regulation by membrane composition and protein-protein interactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
42490
1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine, ≥99.0% (10 mg phospholipid per ml CHCl3, TLC)
C41H78NO8P
54008
1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine, ≥98.0% (TLC)
C41H78NO8P
76548
1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine, ≥97.0% (TLC)
C41H78NO8P
HPA005437
Anti-BIN1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA003894
Anti-BIN1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
S4500
L-Serine, ReagentPlus®, ≥99% (HPLC)
C3H7NO3
RES0908S-A7
L-Serine, PharmaGrade, Ajinomoto, EP, USP, JP, Manufactured under appropriate controls for use in pharma or biopharmaceutical production, suitable for cell culture
C3H7NO3
S4311
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
C3H7NO3
84959
L-Serine, BioUltra, ≥99.5% (NT)
C3H7NO3
54763
L-Serine, certified reference material, TraceCERT®
C3H7NO3
84960
L-Serine, ≥99.0% (NT)
C3H7NO3
78682
L-Serine, tested according to Ph.Eur.
C3H7NO3
PHR1103
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
C3H7NO3
S1315
L-Serine, PharmaGrade, Ajinomoto, EP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C3H7NO3