EMAIL THIS PAGE TO A FRIEND
Molecular nutrition & food research
Antioxidative stress effect of phosphoserine dimers is mediated via activation of the Nrf2 signaling pathway.
PMID 25351664
Abstract
Phosphoserine-containing peptides have been shown to exert antioxidative stress effects, by lowering lipid peroxidation, increasing intracellular glutathione, and increasing the expression of antioxidant enzymes in human intestinal epithelial cells. However, the role of phosphoserine residues in antioxidative stress activity, and their mechanism of action, remains unknown. The antioxidative stress activity of phosphoserine and phosphoserine peptides was examined using an in vitro model of hydrogen peroxide (H2 O2 )-induced oxidative stress in Caco-2 cells. Phosphoserine dimers (2PS) reduced IL-8 secretion in H2 O2 -treated Caco-2 cells, and reduced H2 O2 -induced expression of genes involved in inflammation and generation of reactive oxygen species (ROS), including chemokine (C-C motif) ligand 5 (CCL5), lactoperoxidase (LPO), myeloperoxidase (MPO), neutrophil cytosolic factor 1/2 (NCF1/2), and nitric oxide synthase 2A (NOS2), and upregulated metallothionein 3 (MT3), peroxiredoxin 3 (PRDX3), and surfactant, pulmonary-associated protein D (SFTPD), which are involved in protection against oxidative stress and activation of the Nrf2 signaling pathway. At the protein level, 2PS reduced H2 O2 -induced phosphorylation of the ERK1/2 and JNK MAPKs, and increased Nrf2 expression. Moreover, the ability of 2PS to reduce H2 O2 -induced IL-8 secretion, a marker of inflammation and oxidative stress, was abrogated in Nrf2 knockdown cells. These results suggest that 2PS reduce H2 O2 -induced oxidative stress via the Nrf2 signaling pathway, and reveal a potential mechanism for the antioxidative stress activity of phosphoserine-containing peptides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0878
O-Phospho-L-serine
C3H8NO6P