EMAIL THIS PAGE TO A FRIEND
Clinical oral investigations
Nitric oxide-mediated cytotoxic effect induced by zoledronic acid treatment on human gingival fibroblasts.
PMID 25352469
Abstract
The aim of our work was to evaluate the role of nitric oxide (NO) in the in vitro response of human gingival fibroblasts (HGFs) to 1, 5, 10, and 100xa0μM doses of zoledronic acid (ZA), a bisphosphonate largely used in the clinical practice and for which several adverse effects are reported. Phase contrast microscopy and live/dead staining were used to evaluate HGFs morphology; cell viability, collagen type I and interleukin 6 (IL-6) secretion were evaluated by 3-[4,5-dimethyl-thiazol-2-yl-]-2,5-diphenyl tetrazolium bromide (MTT) and enzyme-linked immunosorbent assay (ELISA) assays. Reactive oxygen species (ROS) production and mitochondrial membrane potential were evaluated by flow cytometry; NO production and NOS activity by spectrophotometric analysis; endothelial NOS (eNOS) and neuronal NOS (nNOS) expression by immunofluorescence. Viable fibroblasts are evidenced in control sample while floating dead cells and cells close to detachment phase in ZA-treated sample, in agreement with decreased level of collagen type I. Control sample shows higher number of viable cells respect to ZA-treated one and ROS production increases when ZA is added. Released NO in ZA-treated sample appears higher and NO overproduction is related to increased nNOS activity. IL-6 secretion level is higher in ZA-treated sample than in control one. Our results suggest ROS involvement in NO overproduction, due to nNOS recruitment, both at low and high doses. In turn, NO release seems to be able to trigger the inflammatory response only when high doses are administered, thus confirming the ZA cytotoxic effect on HGFs. The knowledge of ZA-mediated cytotoxicity mechanisms on HGFs allows to better understand drug pharmacological activity.