EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Copper(I)-catalyzed three-component reaction of terminal propargyl alcohols, aldehydes, and amines: synthesis of 3-amino-2-pyrones and 2,5-dihydrofurans.
PMID 25353157
Abstract
A novel copper(I)-catalyzed three-component reaction for the efficient synthesis of 3-amino-2-pyrones and 2,5-dihydrofurans from propargyl alcohols, aldehydes, and amines has been developed. The starting materials are easily available and the scope of this method is broad. Through mechanistic studies, it is believed that the three-component reaction consists of an A(3) -coupling to propargylic amine, alkyne-allene isomerization, and intramolecular cyclization of the allenol to form a furan. In case of using ethyl glyoxalate as the aldehyde, a ring-opening, lactonization, and isomerization process affords the 3-amino-2-pyrones.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
294985
Allene, ≥95%
C3H4