EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Annexin A2 regulates TRPA1-dependent nociception.
PMID 25355205
Abstract
The transient receptor potential A1 (TRPA1) channel is essential for vertebrate pain. Even though TRPA1 activation by ligands has been studied extensively, the molecular machinery regulating TRPA1 is only poorly understood. Using an unbiased proteomics-based approach we uncovered the physical association of Annexin A2 (AnxA2) with native TRPA1 in mouse sensory neurons. AnxA2 is enriched in a subpopulation of sensory neurons and coexpressed with TRPA1. Furthermore, we observe an increase of TRPA1 membrane levels in cultured sensory neurons from AnxA2-deficient mice. This is reflected by our calcium imaging experiments revealing higher responsiveness upon TRPA1 activation in AnxA2-deficient neurons. In vivo these findings are associated with enhanced nocifensive behaviors specifically in TRPA1-dependent paradigms of acute and inflammatory pain, while heat and mechanical sensitivity as well as TRPV1-mediated pain are preserved in AnxA2-deficient mice. Our results support a model whereby AnxA2 limits the availability of TRPA1 channels to regulate nociceptive signaling in vertebrates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2610
Anti-ANXA2 (291-305) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
AV36588
Anti-ANXA2 antibody produced in rabbit, IgG fraction of antiserum
SAB1410612
Anti-ANXA2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA026630
Anti-TRPA1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105082
Anti-TRPA1 antibody produced in rabbit, affinity isolated antibody
SAB1411593
Anti-TRPA1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB3501027
Anti-TRPV1 antibody produced in rabbit, affinity isolated antibody
L4632
Lithium dodecyl sulfate, ≥98.5% (GC)
C12H25LiO4S
L9781
Lithium dodecyl sulfate, BioReagent, for molecular biology, suitable for electrophoresis
C12H25LiO4S
L5901
Lithium dodecyl sulfate, BioXtra, ≥98.5% (GC)
C12H25LiO4S
WH0000302M1
Monoclonal Anti-ANXA2 antibody produced in mouse, clone 3E8-B6, purified immunoglobulin, buffered aqueous solution
SAB1409173
Monoclonal Anti-ANXA2 antibody produced in mouse, clone 4G8, purified immunoglobulin, buffered aqueous solution
WH0007442M1
Monoclonal Anti-TRPV1 antibody produced in mouse, clone 1F5, purified immunoglobulin, buffered aqueous solution
SAB1404527
Monoclonal Anti-TRPV1 antibody produced in mouse, clone 1A8, purified immunoglobulin, buffered aqueous solution