EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Interaction of the cell adhesion molecule CHL1 with vitronectin, integrins, and the plasminogen activator inhibitor-2 promotes CHL1-induced neurite outgrowth and neuronal migration.
PMID 25355214
Abstract
The cell adhesion molecule close homolog of L1 (CHL1) plays important functional roles in the developing and adult nervous system. In search of the binding partners that mediate the diverse and sometimes opposing functions of CHL1, the extracellular matrix-associated proteins vitronectin and plasminogen activator inhibitor-2 (PAI-2) were identified as novel CHL1 interaction partners and tested for involvement in CHL1-dependent functions during mouse cerebellar development. CHL1-induced cerebellar neurite outgrowth and cell migration at postnatal days 6-8 were inhibited by a CHL1-derived peptide comprising the integrin binding RGD motif, and by antibodies against vitronectin or several integrins, indicating a vitronectin-dependent integrin-mediated pathway. A PAI-2-derived peptide, or antibodies against PAI-2, urokinase type plasminogen activator (uPA), uPA receptor, and several integrins reduced cell migration. CHL1 colocalized with vitronectin, PAI-2, and several integrins in cerebellar granule cells, suggesting an association among these proteins. Interestingly, at the slightly earlier age of 4-5 d, cerebellar neurons did not depend on CHL1 for neuritogenesis and cell migration. However, differentiation of progenitor cells into neurons at this stage was dependent on homophilic CHL1-CHL1 interactions. These observations indicate that homophilic CHL1 trans-interactions regulate differentiation of neuronal progenitor cells at early postnatal stages, while heterophilic trans-interactions of CHL1 with vitronectin, integrins, and the plasminogen activator system regulate neuritogenesis and neuronal cell migration at a later postnatal stage of cerebellar morphogenesis. Thus, within very narrow time windows in postnatal cerebellar development, distinct types of molecular interactions mediated by CHL1 underlie the diverse functions of this protein.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA003345
Anti-CHL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408184
Anti-CHRDL1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA000226
Anti-CHRDL1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA000250
Anti-CHRDL1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408696
Anti-CHRDL1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1306935
ANTI-DDX11 (CENTER) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1400075
Anti-DDX11 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB1306340
ANTI-PAI2 (CENTER) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1406218
Anti-SERPINB2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA015480
Anti-SERPINB2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104454
Anti-SERPINB2 antibody produced in rabbit, affinity isolated antibody
SAB1410870
Anti-SERPINB2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
WH0091851M1
Monoclonal Anti-CHRDL1 antibody produced in mouse, clone 1G11, purified immunoglobulin, buffered aqueous solution
SAB1401112
Monoclonal Anti-DDX11 antibody produced in mouse, clone 3C1, purified immunoglobulin, buffered aqueous solution
WH0005055M8
Monoclonal Anti-SERPINB2 antibody produced in mouse, clone 3A9, purified immunoglobulin, buffered aqueous solution