EMAIL THIS PAGE TO A FRIEND
Scandinavian journal of immunology
Lipocalin-2 promotes m1 macrophages polarization in a mouse cardiac ischaemia-reperfusion injury model.
PMID 25359467
Abstract
Ischaemia-reperfusion (IR) injury is a major issue in cardiac transplantation. Inflammatory processes play a major role in myocardial IR injury. Lipocalin-2 (Lcn2), which is also known as neutrophil gelatinase-associated lipocalin, has multiple functions that include the regulation of cell death/survival, cell migration/invasion, cell differentiation and iron delivery. In our study, the hearts of C57BL/6 mice were flushed with and stored in cold Bretschneider solution for 8xa0h and then transplanted into a syngeneic recipient. We found that Lcn2 neutralization decreased the recruitment of neutrophils and macrophages. Troponin T (TnT) production, 24xa0h after myocardial IR injury, was reduced through anti-Lcn2 antibody administration. The cardiac output at 60xa0mmHg of afterload pressure was significantly increased in hearts administrated with anti-Lcn2 antibody administration (anti-Lcn-2: 58.9xa0±xa05.62xa0ml/min; control: 25.8xa0±xa04.1xa0ml/min; Pxa0