EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Heart and circulatory physiology
Age-related changes in rat dermal extracellular matrix composition affect the distribution of plasma proteins as a function of size and charge.
PMID 25362136
Abstract
Collagen and glycosaminoglycans (GAGs) constituting the ECM may limit the space available and thus exclude macromolecules from a fraction of the interstitial fluid (IF) phase. This exclusion phenomenon is of importance for transcapillary fluid and solute exchange. The purpose of the study was to examine the range of interstitial exclusion in rat skin by using probes within a span of molecular weights and electrical charge and also to test if a change in interstitial composition, occurring as a consequence of aging, affected exclusion. To this end, we used a novel approach, involving the exact determination of albumin concentration and mass in IF and tissue eluate by HPLC and thereafter, expressing the corresponding numbers relative to albumin for a set of probe proteins assessed by quantitative proteomics. Albumin was excluded from 55±4% (n=8) of the extracellular fluid phase. There was a highly significant, positive correlation between probe Stokes-Einstein (SE) radius and fractional excluded volume (VEF), described by VEF=0.078·SE radius+0.269 (P<0.001), and oppositely, a negative correlation between probe isoelectric point (pI) and exclusion for proteins with comparable size, VEF=-0.036·pI+0.719 (P=0.04). Aging resulted in a significant reduction in skin hydration and sulfated GAGs, a moderate increase in hyaluronan, and a corresponding, reduced VEF for albumin and the other macromolecular probes. Our findings suggest that the changes in the ECM in aged skin may result in delayed adjustments of fluid perturbations and reduced ability for salt storage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4387
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
C7806 Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
C1188 Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
C8919 Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
C9791
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
C3511
Collagen from calf skin, Bornstein and Traub Type I (Sigma Type III), solid
C9301 Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
C0543 Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder (from 0.2 μm-filtered solution), BioReagent, suitable for cell culture
C4407 Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
C3657 Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
C8374
Collagen from human placenta, Bornstein and Traub Type IV, powder
H4417 Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, ≥95% (SDS-PAGE)
C3929 Collagen from kangaroo tail, Bornstein and Traub Type I, powder
C5608 Collagen from rabbit skin, Bornstein and Traub Type I, powder
27666 Collagen from rat tail, for cell biology, soluble
C7624
Collagen human, Bornstein and Traub Type I, recombinant, expressed in Nicotiana tabacum (tobacco)
C7999
Collagen human, Bornstein and Traub Type I, recombinant, expressed in Nicotiana tabacum (tobacco)
C8993 Collagen ovine, aqueous solution
C6745 Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
H7630
Hyaluronic acid sodium salt from bovine vitreous humor
H1876
Hyaluronic acid sodium salt from human umbilical cord
H5388
Hyaluronic acid sodium salt from rooster comb, avian glycosaminoglycan polysaccharide
53747
Hyaluronic acid sodium salt from Streptococcus equi, bacterial glycosaminoglycan polysaccharide
49775
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 1,200
40583
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 8,000-15,000
94137
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 10,000-30,000
97616
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 15,000-30,000
42686
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 30,000-50,000
08185
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 50,000-70,000
75048
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 70,000-90,000