EMAIL THIS PAGE TO A FRIEND
Nature chemical biology
Diels-Alder reaction-triggered bioorthogonal protein decaging in living cells.
PMID 25362360
Abstract
Small molecules that specifically activate an intracellular protein of interest are highly desirable. A generally applicable strategy, however, remains elusive. Herein we describe a small molecule-triggered bioorthogonal protein decaging technique that relies on the inverse electron-demand Diels-Alder reaction for eliminating a chemically caged protein side chain within living cells. This method permits the efficient activation of a given protein (for example, an enzyme) in its native cellular context within minutes.