EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Spectroscopic investigation for the interaction of mycophenolate mofetil with ferrous ions.
PMID 25366315
Abstract
The interaction of mycophenolate mofetil (MMF) with ferrous ions (Fe(2+)) in the solid state, in water, and in polar organic solvents was investigated using (1)H-NMR, (13)C-NMR, IR, and UV-visible (Vis) spectroscopies. A red-purple colored substance was formed after grinding solid MMF and FeSO4·7H2O in a mortar. The IR spectrum of taken as a KBr tablet of the colored substance showed a new absorption band at 1651 cm(-1). Although the color disappeared when the sample was dissolved in water, it persisted in organic solvents such as MeOH or dimethyl sulfoxide (DMSO). The UV-Vis spectrum of a 0.25 mM MeOH solution of MMF showed a new absorption maximum at 507 nm in the presence of Fe(2+) ions, while an aqueous solution of the same mixture showed no significant change from the MMF solution. All the signals in the (13)C-NMR spectrum in DMSO-d6 solution were unambiguously assigned. Upon the addition of 0.5 eq. of Fe(2+) ions, all the carbon signals except those of the 2-morpholinoethyl group almost disappeared, which clearly indicated that the Fe(2+) ions were located far away from the 2-morpholinoethyl groups in the MMF molecules. On the basis of these results, we have concluded that the MMF-Fe(2+) complex is actually formed in the solid state as well as in polar organic solvents such as MeOH or DMSO.