EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Active food packaging based on molecularly imprinted polymers: study of the release kinetics of ferulic acid.
PMID 25369799
Abstract
A novel active packaging based on molecularly imprinted polymer (MIP) was developed for the controlled release of ferulic acid. The release kinetics of ferulic acid from the active system to food simulants (10, 20, and 50% ethanol (v/v), 3% acetic acid (w/v), and vegetable oil), substitutes (95% ethanol (v/v) and isooctane), and real food samples at different temperatures were studied. The key parameters of the diffusion process were calculated by using a mathematical modeling based on Fick's second law. The ferulic acid release was affected by the temperature as well as the percentage of ethanol of the simulant. The fastest release occurred in 95% ethanol (v/v) at 20 °C. The diffusion coefficients (D) obtained ranged between 1.8 × 10(-11) and 4.2 × 10(-9) cm(2)/s. A very good correlation between experimental and estimated data was obtained, and consequently the model could be used to predict the release of ferulic acid into food simulants and real food samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
360066
2,2,4-Trimethylpentane, anhydrous, 99.8%
C8H18
650439
2,2,4-Trimethylpentane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.5%
C8H18
34862
2,2,4-Trimethylpentane, for HPLC, ≥99%
C8H18
360597
2,2,4-Trimethylpentane, ACS reagent, ≥99.0%
C8H18
59030
2,2,4-Trimethylpentane, analytical standard
C8H18
59045
2,2,4-Trimethylpentane, puriss. p.a., ACS reagent, ≥99.5% (GC)
C8H18
32291
2,2,4-Trimethylpentane, puriss. p.a., ≥99.5% (GC)
C8H18
15-2200
2,2,4-Trimethylpentane, SAJ first grade, ≥98.0%
C8H18
15-2190
2,2,4-Trimethylpentane, JIS special grade
C8H18
15-2211
2,2,4-Trimethylpentane, for HPLC
C8H18
258776
2,2,4-Trimethylpentane, ReagentPlus®, ≥99%
C8H18
234907
2-(Dimethylamino)ethyl methacrylate, contains 700-1000 ppm monomethyl ether hydroquinone as inhibitor, 98%
C8H15NO2
Y0001489
2-(Dimethylamino)ethyl methacrylate, European Pharmacopoeia (EP) Reference Standard
C8H15NO2
44964
Density Standard 692 kg/m3, H&D Fitzgerald Ltd. Quality
C8H18
40136
Dichlorodimethylsilane, Selectophore
C2H6Cl2Si
440272
Dichlorodimethylsilane, ≥99.5%
C2H6Cl2Si
40140
Dichlorodimethylsilane, ≥98.5% (GC)
C2H6Cl2Si
80430
Dichlorodimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
C2H6Cl2Si
Y0001013
Ferulic acid, European Pharmacopoeia (EP) Reference Standard
C10H10O4
85126
Silanization solution I, ~5% (dimethyldichlorosilane in heptane), Selectophore
C2H6Cl2Si
46278
trans-Ferulic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
C10H10O4
52229
trans-Ferulic acid, certified reference material, TraceCERT®
C10H10O4