EMAIL THIS PAGE TO A FRIEND
Molecular medicine reports
Effects of antiviral therapy on the cellular immune response in patients with chronic hepatitis B.
PMID 25371014
Abstract
A weak T‑cell immune response to the hepatitis B virus (HBV) is hypothesized to be the primary cause of chronic HBV infection. Emerging evidence suggests that long‑term effective antiviral therapy restores the HBV‑specific T‑cell response from exhaustion. However, the extent to which the cellular immune response can be restored following the persistent suppression of HBV replication by antiviral therapy remains unclear. In order to investigate this question, 46 patients with chronic hepatitis B (CHB) treated with nucleos(t)ide analogues who demonstrated persistent suppression of HBV replication [defined as undetectable HBV DNA, hepatitis B e antigen (HBeAg) negative and adherence to antiviral therapy], 22 untreated CHB patients, 15 patients with acute hepatitis B (AHB) and 10 healthy adults were recruited. HBV‑specific interferon‑γ enzyme‑linked immunospot (IFN‑γ ELISPOT) assay and HBV‑specific T‑cell proliferation analysis were performed with a panel of overlapping peptides covering the envelope and core antigens. Data from this study showed that the HBV‑specific immune responses to the peptide pools of the envelope and core protein in the treated patients were stronger than those in the untreated CHB patients, but significantly weaker than those in the AHB patients and healthy adults. A higher frequency of response to S than C peptide pools was confirmed by the IFN‑γ ELISPOT assay in the treated CHB patients. The restoration of antiviral immunity was clearly associated with a reduction in HBV DNA and the duration of HBV DNA suppression. In conclusion, the HBV‑specific immune responses in the CHB patients can be significantly restored from exhaustion following the persistent suppression of HBV replication as a result of antiviral treatment with nucleos(t)ide analogues.