EMAIL THIS PAGE TO A FRIEND
Scientific reports
Quantification of neurotoxin BMAA (β-N-methylamino-L-alanine) in seafood from Swedish markets.
PMID 25373604
Abstract
The neurotoxin β-N-methylamino-L-alanine (BMAA) produced naturally by cyanobacteria, diatoms and dinoflagellates can be transferred and accumulated up the food chain, and may be a risk factor for neurodegenerative diseases. This study provides the first systematic screening of BMAA exposure of a large population through the consumption of seafood sold in metropolitan markets. BMAA was distinguished from known isomers by liquid chromatography tandem mass spectrometry after acidic hydrolysis and derivatization. Using deuterium-labeled internal standard, BMAA was quantified as 0.01-0.90 μg/g wet weight of tissues in blue mussel, oyster, shrimp, plaice, char and herring, but was undetectable (<0.01 μg/g) in other samples (salmon, cod, perch and crayfish). Provided that the content of BMAA detected is relevant for intake calculations, the data presented may be used for a first estimation of BMAA exposure through seafood from Swedish markets, and to refine the design of future toxicological experiments and assessments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32830
L-2,4-Diaminobutyric acid dihydrochloride, ≥95.0%
C4H10N2O2 · 2HCl