EMAIL THIS PAGE TO A FRIEND
International ophthalmology
Comparison of efficacy of three surgical methods of conjunctival autograft fixation in the treatment of pterygium.
PMID 25374052
Abstract
Our aim primarily was to compare the recurrence rate with three techniques of conjunctival fixation (suture versus fibrin glue versus autologous in situ blood coagulum) over bare sclera following pterygium excision. Ninety eyes of 90 patients with primary pterygium were randomly divided into three groups: group I (30 eyes) underwent autografting and fixation with 8-0 vicryl sutures, group II (30 eyes) with fibrin glue and group III (30 eyes) with autologous in situ blood coagulum. The patients were reviewed on 2nd day, weeks 1 and 4, and at every 3 months till 12 months after surgery. Rate of recurrence was similar (p = 0.585) across the three groups. Time taken for surgery for Group 1 was more as compared to group 2 (p < 0.001) and group 3 (p < 0.001). Also, group 2 cases took significantly more time as compared to group 3 (p < 0.001). Postoperative patient discomfort (foreign body sensation, epiphora, pain and irritation) was more in suture-assisted autografting as compared to the other two groups. However, at some points along the time line, patient discomfort was significantly more in group III as compared to group II. Complications like graft retraction, graft displacement and cyst formation were seen in a few patients but were not statistically significant across the three groups. All three techniques were found to be useful methods and were associated with similar rate of recurrence.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1756
Dexamethasone, ≥98% (HPLC), powder
C22H29FO5
D9184
Dexamethasone, meets USP testing specifications
C22H29FO5
D0700000
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
C22H29FO5
1176007
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
C22H29FO5
46165
Dexamethasone, VETRANAL, analytical standard
C22H29FO5
31375
Dexamethasone, tested according to Ph.Eur.
C22H29FO5
PHR1526
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
C22H29FO5
Y0001593
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
C22H29FO5
Y0001177
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
C22H29FO5
O8757
Ofloxacin, fluoroquinolone antibiotic
C18H20FN3O4
33703
Ofloxacin, VETRANAL, analytical standard
C18H20FN3O4
1478108
Ofloxacin, United States Pharmacopeia (USP) Reference Standard
C18H20FN3O4
PHR1168 Ofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
C18H20FN3O4
P8650
Prednisolone 21-acetate, ≥97%
C23H30O6
46657
Prednisolone 21-acetate, VETRANAL, analytical standard
C23H30O6
P2800000
Prednisolone acetate, European Pharmacopoeia (EP) Reference Standard
C23H30O6
1556008
Prednisolone acetate, United States Pharmacopeia (USP) Reference Standard
C23H30O6
Y0000582
Prednisolone acetate for peak identification, European Pharmacopoeia (EP) Reference Standard
C23H30O6