PloS one
Eribulin mesylate targets human telomerase reverse transcriptase in ovarian cancer cells.
PMID 25375122
Abstract
Treatment of advanced ovarian cancer involves platinum-based chemotherapy. However, chemoresistance is a major obstacle. Cancer stem cells (CSCs) are thought to be one of the causes of chemoresistance, but the underlying mechanism remains elusive. Recently, human telomerase reverse transcriptase (hTERT) has been reported to promote CSC-like traits. In this study, we found that a mitotic inhibitor, eribulin mesylate (eribulin), effectively inhibited growth of platinum-resistant ovarian cancer cell lines. Eribulin-sensitive cells showed a higher efficiency for sphere formation, suggesting that these cells possess an enhanced CSC-like phenotype. Moreover, these cells expressed a higher level of hTERT, and suppression of hTERT expression by siRNA resulted in decreased sensitivity to eribulin, suggesting that hTERT may be a target for eribulin. Indeed, we found that eribulin directly inhibited RNA-dependent RNA polymerase (RdRP) activity, but not telomerase activity of hTERT in vitro. We propose that eribulin targets the RdRP activity of hTERT and may be an effective therapeutic option for CSCs. Furthermore, hTERT may be a useful biomarker to predict clinical responses to eribulin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1407312
Anti-GNL3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1412580
ANTI-GNL3 antibody produced in mouse, clone 1G4, purified immunoglobulin, buffered aqueous solution
SAB1412581
ANTI-GNL3 antibody produced in mouse, clone 4B10, purified immunoglobulin, buffered aqueous solution
SAB1412582
ANTI-GNL3 antibody produced in mouse, clone 2B9, purified immunoglobulin, buffered aqueous solution
SAB2106203
Anti-GNL3 antibody produced in rabbit, affinity isolated antibody
SAB2104519
Anti-GNL3, (N-terminal) antibody produced in rabbit, affinity isolated antibody
GW22621 Anti-Nucleostemin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
HPA009067
Anti-SPINK5 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA011351
Anti-SPINK5 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1307204
ANTI-SPINK5(N-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4200195
Monoclonal Anti-Brg1 antibody produced in rat, ~1.0 mg/mL, clone 5B7, purified immunoglobulin
WH0026354M1
Monoclonal Anti-GNL3 antibody produced in mouse, clone 1A1, purified immunoglobulin, buffered aqueous solution
SAB1405018
Monoclonal Anti-GNL3 antibody produced in mouse, clone 3A3, purified immunoglobulin, buffered aqueous solution