PloS one
Effects of sex, age, and fasting conditions on plasma lipidomic profiles of fasted Sprague-Dawley rats.
PMID 25375860
Abstract
Circulating lipid molecules reflect biological processes in the body and, thus, are useful tools for preclinical estimation of the efficacy and safety of newly developed drugs. However, background information on profiles of circulating lipid molecules in preclinical animal models is limited. Therefore, we examined the effects of multiple factors such as sex (fasted male vs. female), age (fasted 10 vs. 30 weeks old), and feeding conditions (feeding vs. fasting, 16 vs. 22 hr fasting, 10 AM vs. 4 PM blood collection), on the global profiles of lipid molecules in plasma from Sprague-Dawley rats by using a lipidomic approach. Our assay platform determined 262 lipid molecules (68 phospholipids, 20 sphingolipids, 138 neutral lipids, and 36 polyunsaturated fatty acids and their metabolites) in rat plasma. Multivariate discriminant analysis (orthogonal partial least squares discriminant analysis) and heat maps of statistically significant lipid molecules revealed that the plasma lipid profiles in rats are predominantly influenced by feeding conditions, followed by sex and age. In addition, the fasting duration (16 vs. 22 hr fasting) or the time of blood collection (10 AM vs. 4 PM blood collection) has limited or no contribution on the profiles of lipid molecules in rat plasma. Our results provide useful, fundamental information for exploring and validating biomarkers in future preclinical studies and may help to establish regulatory standards for such studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L0517
Leukotriene B4, ~100 μg/mL in ethanol, ≥97%
C20H32O4