EMAIL THIS PAGE TO A FRIEND
Psychiatria Danubina
Treatment of insomnia with hypnotics resulting in improved sexual functioning in post-menopausal women.
PMID 25377370
Abstract
This study sought to determine whether trazodone used in hypnotic doses, compared to the hypnotic agent zopiclone, had any specific positive effect on sexual function in non-depressive post-menopausal women with insomnia. Fifty (50) subjects participated in the study. Insomnia and sexual performance were evaluated before and after 4 weeks of hypnotic treatment. At week four, both treatments improved sleep quality to a similar degree. Sexual function also improved significantly with both treatments, with no significant difference between the groups. In post-menopausal women, sexual problems and sleep problems may be related and solving sleep problems may help sexual functioning, independently of depression.