EMAIL THIS PAGE TO A FRIEND
Japanese journal of clinical oncology
Randomized controlled trial comparing docetaxel-cisplatin combination with weekly docetaxel alone in elderly patients with advanced non-small-cell lung cancer: Japan Clinical Oncology Group (JCOG) 0207†.
PMID 25378648
Abstract
Prospective trials specifically designed for elderly patients with advanced non-small-cell lung cancer demonstrating the benefit of platinum-based therapies are still lacking. This trial was designed to clarify whether the addition of cisplatin to monotherapy could improve survival for elderly patients. Elderly patients (age ≥70 years, ECOG performance Status 0-1) with advanced non-small-cell lung cancer were randomized to receive docetaxel 20 mg/m(2) plus cisplatin 25 mg/m(2) on Day 1, 8 and 15 (docetaxel plus cisplatin) or docetaxel 25 mg/m(2) on the same schedule (docetaxel). Both regimens were repeated every 4 weeks until disease progression. One hundred and twenty-six patients were enrolled. Sixty-three were randomly assigned docetaxel plus cisplatin and 63 docetaxel monotherapy. Median age was 76 years (range 70-88). The second planned interim analysis was performed on 112 assessable patients (docetaxel/docetaxel plus cisplatin: 56/56). Although the formal criterion for stopping the trial was not met, the Data and Safety Monitoring Committee recommended study termination on ethical grounds based on the interaction (two-sided P = 0.077, hazard ratios for ≤74/≥75: 0.23/0.72) between age and subgroup and treatment arm, which suggested that docetaxel may not represent an adequate control arm regimen for the age subgroup of 70-74 years. The interpretation of study results is limited due to early stopping. Further study is needed to confirm survival benefit of platinum-based chemotherapy for elderly non-small-cell lung cancer [UMIN-CTR (www.umin.ac.jp/ctr/) ID: C000000146].