EMAIL THIS PAGE TO A FRIEND
Zebrafish
The tumor suppressor rpl36 restrains KRAS(G12V)-induced pancreatic cancer.
PMID 25380065
Abstract
Ribosomal proteins are known to be required for proper assembly of mature ribosomes. Recent studies indicate an additional role for ribosomal proteins as candidate tumor suppressor genes. Pancreatic acinar cells, recently identified as effective cells of origin for pancreatic adenocarcinoma, display especially high-level expression of multiple ribosomal proteins. We, therefore, functionally interrogated the ability of two ribosomal proteins, rpl36 and rpl23a, to alter the response to oncogenic Kras in pancreatic acinar cells using a newly established model of zebrafish pancreatic cancer. These studies reveal that rpl36, but not rpl23a, acts as a haploinsufficient tumor suppressor, as manifested by more rapid tumor progression and decreased survival in rpl36(hi1807/+);ptf1a:gal4VP16(Tg);UAS:GFP-KRAS(G12V) fish compared with their rpl36(+/+);ptf1a:gal4VP16;UAS:GFP-KRAS(G12V) siblings. These results suggest that rpl36 may function as an effective tumor suppressor during pancreatic tumorigenesis.