EMAIL THIS PAGE TO A FRIEND
Journal of gastroenterology
Overexpression of NRG1 promotes progression of gastric cancer by regulating the self-renewal of cancer stem cells.
PMID 25381017
Abstract
Gastric cancer stem cells (GCSCs) have been successfully isolated from patients. However, the molecular mechanisms underlying the self-renewal of GCSCs and their relationship with the microenvironment are poorly characterized. GCSCs and cancer-associated fibroblasts (CAFs) were cultured directly from gastric cancer patients. The self-renewal of GCSCs was assayed by sphere formation assay and in vivo tumorigenicity. Expression of neuregulin1 (NRG1) was examined by immunohistochemistry, real-time PCR and western blotting. CAFs increased the self-renewal of GCSCs by secreting NRG1. NRG1 activated NF-κB signaling and this activation regulated GCSC self-renewal. Moreover, NF-κB-active GCSCs were tumorigenic, however NF-κB-inactive GCSCs were not. The overexpression of NRG1 in stromal cells and cancer cells was observed in the tumor tissues of gastric cancer patients and was associated with clinical stage lymph node metastasis and survival in gastric cancer patients. In addition, we also found that NRG1 can regulate the proliferation and invasion of gastric cancer cells. These results indicate that NRG1, which can be secreted by CAFs or cancer cells, promotes progression of gastric cancer by regulating the self-renewal of GCSCs and its overexpression is associated with a prognosis of gastric cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300579
Anti-ERBB3 (Ab-1289) antibody produced in rabbit, affinity isolated antibody
SAB4300580
Anti-ERBB3 (Ab-1328) antibody produced in rabbit, affinity isolated antibody
HPA045396
Anti-ERBB3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA012016
Anti-ERBB4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500790
Anti-HER3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500791
Anti-HER3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500792
Anti-HER3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1305670
ANTI-HER4 (C-TERM Y1162) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4500794
Anti-HER4, C-Terminal antibody produced in rabbit, affinity isolated antibody
HPA010964
Anti-NRG1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV46957
Anti-NRG1 antibody produced in rabbit, affinity isolated antibody
SAB2101655
Anti-NRG1 antibody produced in rabbit, affinity isolated antibody
SAB1410627
Anti-NRG1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4300058
Anti-phospho-ERBB2 (pTyr1221/pTyr1222) antibody produced in rabbit, affinity isolated antibody
SAB4300061
Anti-phospho-ERBB2 (pTyr1248) antibody produced in rabbit, affinity isolated antibody
SAB4300057
Anti-phospho-ERBB2 (pTyr877) antibody produced in rabbit, affinity isolated antibody
SAB4300265
Anti-phospho-ERBB3 (pTyr1328) antibody produced in rabbit, affinity isolated antibody
SAB4504586
Anti-phospho-HER2 (pTyr1112) antibody produced in rabbit, affinity isolated antibody
SAB4504425
Anti-phospho-HER2 (pTyr1221/Tyr1222) antibody produced in rabbit, affinity isolated antibody
SAB4504427
Anti-phospho-HER2 (pTyr877) antibody produced in rabbit, affinity isolated antibody
SAB1305460 ANTI-PHOSPHO-HER2(Y1112) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4504741
Anti-phospho-HER3 (pTyr1197) antibody produced in rabbit, affinity isolated antibody
SAB4504193
Anti-phospho-HER3 (pTyr1222) antibody produced in rabbit, affinity isolated antibody
SAB4504194
Anti-phospho-HER3 (pTyr1289) antibody produced in rabbit, affinity isolated antibody
SAB4504195
Anti-phospho-HER4 (pTyr1284) antibody produced in rabbit, affinity isolated antibody
SAB1306712
ANTI-PHOSPHO-HER4(Y1162) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306304 ANTI-PHOSPHO-HER4(Y1188) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
RAB0388
Human NRG1-β 1 ELISA Kit, for serum, plasma, cell culture supernatant and urine
E5900 Monoclonal Anti-c-erbB-4 antibody produced in mouse, ~2 mg/mL, clone HER4-36, purified immunoglobulin, buffered aqueous solution
WH0002065M1
Monoclonal Anti-ERBB3 antibody produced in mouse, clone 2E9, purified immunoglobulin, buffered aqueous solution