EMAIL THIS PAGE TO A FRIEND
Journal of the National Cancer Institute
DPYD variants as predictors of 5-fluorouracil toxicity in adjuvant colon cancer treatment (NCCTG N0147).
PMID 25381393
Abstract
Previous studies have suggested the potential importance of three DPYD variants (DPYD*2A, D949V, and I560S) with increased 5-FU toxicity. Their individual associations, however, in 5-FU-based combination therapies, remain controversial and require further systematic study in a large patient population receiving comparable treatment regimens with uniform clinical data. We genotyped 2886 stage III colon cancer patients treated adjuvantly in a randomized phase III trial with FOLFOX or FOLFIRI, alone or combined with cetuximab, and tested the individual associations between functionally deleterious DPYD variants and toxicity. Logistic regressions were used to assess univariate and multivariable associations. All statistical tests were two-sided. In 2594 patients with complete adverse event (AE) data, the incidence of grade 3 or greater 5FU-AEs in DPYD*2A, I560S, and D949V carriers were 22/25 (88.0%), 2/4 (50.0%), and 22/27 (81.5%), respectively. Statistically significant associations were identified between grade 3 or greater 5FU-AEs and both DPYD*2A (odds ratio [OR] = 15.21, 95% confidence interval [CI] = 4.54 to 50.96, P < .001) and D949V (OR = 9.10, 95% CI = 3.43 to 24.10, P < .001) variants. Statistical significance remained after adjusting for multiple variables. The DPYD*2A variant statistically significantly associated with the specific AEs nausea/vomiting (P = .007) and neutropenia (P < .001), whereas D949V statistically significantly associated with dehydration (P = .02), diarrhea (P = .003), leukopenia (P = .002), neutropenia (P < .001), and thrombocytopenia (P < .001). Although two patients with I560S had grade≥3 5FU-AEs; a statistically significant association could not be demonstrated because of its low frequency (P = .48). In the largest study to date, statistically significant associations were found between DPYD variants (DPYD*2A and D949V) and increased incidence of grade 3 or greater 5FU-AEs in patients treated with adjuvant 5-FU-based combination chemotherapy.