EMAIL THIS PAGE TO A FRIEND
Ecotoxicology (London, England)
In situ effects of pesticides on amphibians in the Sierra Nevada.
PMID 25381462
Abstract
For more than 20xa0years, conservationists have agreed that amphibian populations around the world are declining. Results obtained through laboratory or mesocosm studies and measurement of contaminant concentrations in areas experiencing declines have supported a role of contaminants in these declines. The current study examines the effects of contaminant exposure to amphibians in situ in areas actually experiencing declines. Early larval Pseudacris regilla were translocated among Lassen Volcanic, Yosemite and Sequoia National Parks, California, USA and caged in wetlands in 2001 and 2002 until metamorphosis. Twenty contaminants were identified in tadpoles with an average of 1.3-5.9 (maximumxa0=xa010) contaminants per animal. Sequoia National Park, which had the greatest variety and concentrations of contaminants in 2001, also had tadpoles that experienced the greatest mortality, slowest developmental rates and lowest cholinesterase activities. Yosemite and Sequoia tadpoles and metamorphs had greater genotoxicity than those in Lassen during 2001, as determined by flow cytometry. In 2001 tadpoles at Yosemite had a significantly higher rate of malformations, characterized as hemimelia (shortened femurs), than those at the other two parks but no significant differences were observed in 2002. Fewer differences in contaminant types and concentrations existed among parks during 2002 compared to 2001. In 2002 Sequoia tadpoles had higher mortality and slower developmental rates but there was no difference among parks in cholinesterase activities. Although concentrations of most contaminants were below known lethal concentrations, simultaneous exposure to multiple chemicals and other stressors may have resulted in lethal and sublethal effects.