EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Development of a multi-residue method in a fetal matrix: analysis of meconium.
PMID 25381610
Abstract
Meconium is the earliest stool of newborns. It is a complex matrix that reflects the degree of fetal exposure to environmental pollutants. To investigate exposure to xenobiotics, an analytical method was developed to identify and quantify some pesticides and their metabolites and BTEX metabolites in meconium. Samples were prepared by two liquid-solid extractions and purified twice using SPE cartridges, followed by analysis with liquid chromatography coupled with tandem mass spectrometry. SPE cartridges (polymeric phase with hydrophilic and hydrophobic interactions, ion exchange, mixed mode) were tested and matrix effects were evaluated to determine purification performance. The quantification limits in meconium of this multi-residue method were in the range of 30xa0ngxa0g(-1). The analytical method was applied to "real" meconium samples. Some target analytes were determined in most samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
76514
2,4-D, certified reference material, TraceCERT®
C8H6Cl2O3
31518
2,4-D, PESTANAL®, analytical standard
C8H6Cl2O3
49083
2,4-D, analytical standard
C8H6Cl2O3
D7299
2,4-Dichlorophenoxyacetic acid, plant cell culture tested, ≥98%, crystalline
C8H6Cl2O3
D70724
2,4-Dichlorophenoxyacetic acid, 97%
C8H6Cl2O3
328006
2-Methylhippuric acid, 98%
C10H11NO3
442361
2-Methylphenol, analytical standard
C7H8O
328014
3-Methylhippuric acid, 98%
C10H11NO3
328022
4-Methylhippuric acid, 98%
C10H11NO3
45806
α-Cypermethrin, PESTANAL®, analytical standard
C22H19Cl2NO3
46003
β-Cyfluthrin, PESTANAL®, analytical standard
C22H18Cl2FNO3
33738
Baythroid®, PESTANAL®, analytical standard
C22H18Cl2FNO3
36758
Clopyralid, PESTANAL®, analytical standard
C6H3Cl2NO2
36128
Cypermethrin, PESTANAL®, analytical standard
C22H19Cl2NO3
51991
Cypermethrin, certified reference material, TraceCERT®
C22H19Cl2NO3
05995
Deltamethrin, certified reference material, TraceCERT®
C22H19Br2NO3
45423
Deltamethrin, PESTANAL®, analytical standard
C22H19Br2NO3
D9315
Deltamethrin, ≥98%
C22H19Br2NO3
PS2071
Deltamethrin, analytical standard
C22H19Br2NO3
274623
Dess-Martin periodinane, 97%
C13H13IO8
45428
Diazinon, PESTANAL®, analytical standard
C12H21N2O3PS
68486
Diazinon, certified reference material, TraceCERT®
C12H21N2O3PS
49021
Diazinon, analytical standard
C12H21N2O3PS
45441
Dichlorvos, PESTANAL®, analytical standard
C4H7Cl2O4P
CDS014665
dimethyl phosphate, AldrichCPR
C2H7O4P
M2625
DL-Mandelic acid
C8H8O3
41605
DL-Mandelic acid, analytical reference material
C8H8O3
45521
Glyphosate, PESTANAL®, analytical standard
C3H8NO5P
112003
Hippuric acid, 98%
C9H9NO3
M2101
Mandelic acid, 99%
C8H8O3