EMAIL THIS PAGE TO A FRIEND
Psychopharmacology
Differential effects of clozapine, metoclopramide, haloperidol and risperidone on acquisition and performance of operant responding in rats.
PMID 25381749
Abstract
Prior research has not systematically investigated the effects of systemic antipsychotic drugs on operant response acquisition, specifically their behavioural microstructure, reinforcement blunting and relative potency in acquisition compared to performance once operant responding has stabilized. This study aims to systematically investigate the effects of systemically administered clozapine, metoclopramide, haloperidol and risperidone during free operant response acquisition and performance. Following magazine training, food-restricted male Wistar rats lever pressed for food reward in 15 min daily operant conditioning sessions. All drugs suppressed operant response acquisition and performance. Risperidone and metoclopramide, but not clozapine or haloperidol, suppressed operant responding more potently during acquisition than performance. The dopamine D2-like receptor antagonists haloperidol and metoclopramide that affect the ventral and dorsal striatum blunted reinforcement and decreased inactive lever presses in acquisition. In contrast, the atypical antipsychotics clozapine and risperidone that affect the ventral striatum and prefrontal cortex failed to decrease inactive lever presses during acquisition, suggesting a possible decision-making deficit. Haloperidol decreased active lever pressing over performance days. The drugs did not appear to affect rats' sensitivity to active lever press outcome, even though they suppressed active lever pressing. Results suggest that reinforcement impact during operant acquisition is dependent on dopamine D2 receptors while drugs affecting, among other areas, the prefrontal cortex produce a deficit in ability to suppress inactive lever press responses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6305
Clozapine
C18H19ClN4
C171
Clozapine, solid
C18H19ClN4
C2460000
Clozapine, European Pharmacopoeia (EP) Reference Standard
C18H19ClN4
1142107
Clozapine, United States Pharmacopeia (USP) Reference Standard
C18H19ClN4
Y0000758
Clozapine for peak identification, European Pharmacopoeia (EP) Reference Standard
C18H19ClN4
1142108
Clozapine Resolution Mixture, United States Pharmacopeia (USP) Reference Standard
C18H19ClN4
H1512
Haloperidol, powder
C21H23ClFNO2
1303002
Haloperidol, United States Pharmacopeia (USP) Reference Standard
C21H23ClFNO2
H100
Haloperidol
C21H23ClFNO2
H0100000
Haloperidol, European Pharmacopoeia (EP) Reference Standard
C21H23ClFNO2
Y0001518
Haloperidol for peak identification, European Pharmacopoeia (EP) Reference Standard
C21H23ClFNO2
Y0001517
Haloperidol for system suitability, European Pharmacopoeia (EP) Reference Standard
C21H23ClFNO2
32473
Metoclopramide, VETRANAL, analytical standard
C14H22ClN3O2
M1824990
Metoclopramide, European Pharmacopoeia (EP) Reference Standard
C14H22ClN3O2
R3030
Risperidone, ≥98% (HPLC), powder
C23H27FN4O2
R1000600
Risperidone, European Pharmacopoeia (EP) Reference Standard
C23H27FN4O2
1604654
Risperidone, United States Pharmacopeia (USP) Reference Standard
C23H27FN4O2
Y0000370
Risperidone for system suitability, European Pharmacopoeia (EP) Reference Standard
C23H27FN4O2