EMAIL THIS PAGE TO A FRIEND
Oncogene
Iterative tyrosine phosphorylation controls non-canonical domain utilization in Crk.
PMID 25381819
Abstract
Crk, the prototypical member of a class of Src homology-2 (SH2) and Src homology-3 (SH3) domain containing proteins that controls the coordinated assembly of signaling complexes, is regulated by phosphorylation of Y221 in the linker region, which forms an intramolecular SH2-pY221 auto-clamp to interrupt SH2-N-terminal SH3 domain (SH3N) signaling. Here, we show using LC-MS/MS and by generating phospho-specific antibodies that, iteratively with Y221, the Crk C-terminal SH3 domain (SH3C) is routinely phosphorylated on Y239 and/or Y251 by several extracellular stimuli known to engage Crk. Although phosphorylation at Y221 auto-inhibits the Crk SH2, phosphorylation of the SH3C generates an unconventional phosphoSH3C-SH3N unit in which the SH3N is fully functional to bind polyproline type II ligands and the phosphoSH3C binds de novo to other SH2 domains. Using high-throughput SH2 domain profiling, artificial neural network and position-specific scoring matrix-based bioinformatics approaches, and unbiased mass spectometry, we found that the phosphoSH3C binds several SH2 domain containing proteins, including specific non-receptor tyrosine kinases-Abl via pY251 and C-terminal Src kinase via pY239. Functionally, we show that the phosphoSH3C modulates the Abl-mediated phenotypes of cell spreading and motility. Together, these studies describe a versatile mechanism wherein phosphorylation of Crk at Y221 is not an off switch but redirects signaling from the SH2-SH3N axis to a phosphoSH3C-SH3N axis, with the SH3N as a common denominator.