EMAIL THIS PAGE TO A FRIEND
Inflammation
Overexpression and Potential Regulatory Role of IL-17F in Pathogenesis of Chronic Periodontitis.
PMID 25384562
Abstract
The objective of this study was to investigate the expression level, clinical significance, and possible regulating role of IL-17F in patients of chronic periodontitis. Periodontal local tissues were obtained from chronic periodontitis (CP) and healthy controls (HC) for real-time PCR (RT-PCR) detection with IL-17F and IL-17A messenger RNA (mRNA). Primary human gingival fibroblasts (HGF) were derived from patients receiving crown-lengthening procedures. Efficiency of small interfering RNA (siRNA) of IL-17R to HGF cells were assessed by Western blot and RT-PCR. Recombinant IL-17F and IL-17A were used to stimulate the HGF cells compared with the control group. Aspects of the nuclear factor-kappa B (NF-κB) and extracellular signal-regulated kinase (ERK) signaling pathways were examined by Western blot. Production of pro-inflammatory cytokines induced by IL-17F and IL-17A was detected by RT-PCR. Statistical analysis was analyzed by SPSS software. It showed significantly elevated levels of IL-17F and IL-17A mRNA in CP gingival tissues compared with HC group (P<0.01). Further analysis showed a significant correlation between IL-17F and IL-17A mRNA in CP group (P<0.05), and both cytokines also correlated with the probing depth (P<0.05). Recombinant IL-17F can induce NF-κB phosphor-p65 and ERK phosphorylation of HGF cells similar to that of IL-17A. Interestingly, we found that both IL-17F and IL-17A could promote the important inflammatory cytokines IL-6, CXCL8, and CCL20 production compared with IL-17R siRNA group (P<0.05). This study indicates that IL-17F may be involved in pathogenesis of periodontitis like IL-17A. The role of IL-17F in disease pathogenesis needs to be further investigated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300542
Anti-SYT1 (Ab-309) antibody produced in rabbit, affinity isolated antibody
SAB2501176
Anti-SYT1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA008394
Anti-SYT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA064788
Anti-SYT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107391
Anti-SYT1 antibody produced in rabbit, affinity isolated antibody
SRP3080 IL-17 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
SRP4178 IL-17A from rat, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)
SRP3206 IL-17F from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
SRP6043 IL-17F from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
SRP4176F IL-17F human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
I3525 Interleukin-17A human, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, cell culture tested
H7791 Interleukin-17A human, IL-17A, recombinant, expressed in HEK 293 cells, HumanKine®, cell culture tested
H7916 Interleukin-17F human, HumanKine®, recombinant, expressed in HEK 293 cells, cell culture tested, endotoxin, tested
WH0006857M5
Monoclonal Anti-SYT1 antibody produced in mouse, clone 1A6, purified immunoglobulin, buffered aqueous solution
SAB1404433
Monoclonal Anti-SYT1 antibody produced in mouse, clone 3A5, purified immunoglobulin, buffered aqueous solution
SAB5300361
Monoclonal Anti-SYT1 antibody produced in mouse, clone 8G11B10, ascites fluid