EMAIL THIS PAGE TO A FRIEND
Inflammation
Inflammatory Stress Exacerbated Mesangial Foam Cell Formation and Renal Injury via Disrupting Cellular Cholesterol Homeostasis.
PMID 25387652
Abstract
Inflammation and lipids play significant roles in the progression of chronic kidney disease. This study was designed to investigate whether inflammation disrupts cellular cholesterol homeostasis and causes the lipid nephrotoxicity in vitro and in vivo, and explored its underlying mechanisms. Inflammatory stress was induced by cytokines (interleukin-1β (IL-1β); tumor necrosis factor α (TNF-α)) to human mesangial cells (HMCs) in vitro and by subcutaneous casein injection in C57BL/6J mice in vivo. The data showed that inflammatory stress exacerbated renal cholesterol ester accumulation in vitro and in vivo. Inflammation increased cellular cholesterol uptake and synthesis via upregulating the expression of low-density lipoprotein receptor (LDLr) and 3-hydroxy-3-methylglutaryl-coenzyme A reductase (HMGCoA-R), while it decreased cholesterol efflux via downregulating the expression of liver X receptor alpha and ATP-binding cassette transporter A1. The increased lipid accumulation by inflammatory stress induced reactive oxygen species (ROS) and increased levels of endoplasmic reticulum (ER) stress markers (inositol-requiring protein 1 and activating transcription factor 6) in HMCs and kidneys of C57BL/6J mice. This study implied that inflammation promoted renal lipid accumulation and foam cell formation by disrupting cellular cholesterol homeostasis. Increased intracellular lipids under inflammatory stress caused oxidative stress and ER stress in vitro and in vivo which may contribute to renal injury and progression of chronic kidney disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2106639
Anti-ABCA1 antibody produced in rabbit, affinity isolated antibody
L5044
Anti-LXRα (N-terminal) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
L5169
Anti-LXRβ (C-terminal) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
AV38906
Anti-NR1H2 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV45633
Anti-NR1H2 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB1412663
ANTI-NR1H2 antibody produced in mouse, clone 1B6, purified immunoglobulin, buffered aqueous solution
SAB2103395
Anti-NR1H2 antibody produced in rabbit, affinity isolated antibody
HPA056838
Anti-NR1H2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406923
Anti-NR1H3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV38670
Anti-NR1H3 antibody produced in rabbit, affinity isolated antibody
AV38669
Anti-NR1H3 antibody produced in rabbit, IgG fraction of antiserum
HPA036443
Anti-NR1H3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106276
Anti-NR1H3 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2700416
Anti-SCAP (C-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1303066
ANTI-SCAP (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1303555
ANTI-SCAP (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA004713 Anti-SCAP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406769
Anti-SH2D2A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1409972
Anti-SH2D2A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1401451
Anti-SH2D2A antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4503535
Anti-SH2D2A antibody produced in rabbit, affinity isolated antibody
AV09037
Anti-SREBF2 antibody produced in rabbit, affinity isolated antibody
SAB2102297
Anti-SREBF2 antibody produced in rabbit, affinity isolated antibody
HPA031962 Anti-SREBF2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2103063
Anti-SREBF2 antibody produced in rabbit, affinity isolated antibody
SAB1402677 Monoclonal Anti-ABCA1 antibody produced in mouse, clone 1H4, ascites fluid
WH0007376M1
Monoclonal Anti-NR1H2 antibody produced in mouse, clone 2H2-H3, purified immunoglobulin, buffered aqueous solution
SAB1404516
Monoclonal Anti-NR1H2 antibody produced in mouse, clone 1E1, purified immunoglobulin, buffered aqueous solution
SAB1402495
Monoclonal Anti-NR1H3 antibody produced in mouse, clone 3E9, purified immunoglobulin, buffered aqueous solution
SAB4100468
Monoclonal Anti-SH2D2A antibody produced in mouse, clone SH2D2A-B405, culture supernatant