EMAIL THIS PAGE TO A FRIEND
European journal of pediatrics
Transaldolase deficiency caused by the homozygous p.R192C mutation of the TALDO1 gene in four Emirati patients with considerable phenotypic variability.
PMID 25388407
Abstract
Transaldolase deficiency is a heterogeneous disorder of carbohydrate metabolism characterized clinically by dysmorphic features, cutis laxa, hepatosplenomegaly, hepatic fibrosis, pancytopenia, renal and cardiac abnormalities, and urinary excretion of polyols. This report describes four Emirati patients with transaldolase deficiency caused by the homozygous p.R192C missense mutation in TALDO1 displaying wide phenotypic variability. The patients had variable clinical presentations including hepatosplenomegaly, pancytopenia, liver failure, proteinuria, hydrops fetalis, cardiomyopathy, and skin manifestations (e.g., dryness, cutis laxa, ichthyosis, telangiectasias, and hemangiomas). Biochemical analyses including urinary concentration of polyols were consistent with transaldolase deficiency. The mutation p.R192C was previously identified in an Arab patient, suggesting a founder effect in Arab populations. The above findings support the premise that biallelic mutations in TALDO1 are responsible for transaldolase deficiency and confirm the broad phenotypic variability of this condition, even with the same genotype.