EMAIL THIS PAGE TO A FRIEND
Molecular endocrinology (Baltimore, Md.)
Thermogenic activity of UCP1 in human white fat-derived beige adipocytes.
PMID 25389910
Abstract
Heat-producing beige/brite (brown-in-white) adipocytes in white adipose tissue have the potential to suppress metabolic disease in mice and hold great promise for the treatment of obesity and type 2 diabetes in humans. Here, we demonstrate that human adipose-derived stromal/progenitor cells (hASCs) from subcutaneous white adipose tissue can be efficiently converted into beige adipocytes. Upon pharmacological activation of peroxisome proliferator-activated receptor-γ, hASC-derived adipocytes activated beige fat-selective genes and a brown/beige fat-selective electron transport chain gene program. Importantly, hASC-derived beige fat cells displayed the bioenergetic characteristics of genuine brown fat cells, including a capacity for increased respiratory uncoupling in response to β-adrenergic agonists. Furthermore, knock-down experiments reveal that the thermogenic capacity of human beige fat cells was entirely dependent on the presence of Uncoupling protein 1. In summary, this study reveals that hASCs can be readily differentiated into beige adipocytes that, upon activation, undergo uncoupling protein 1-dependent thermogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I6504
(−)-Isoproterenol hydrochloride
C11H17NO3 · HCl
D199303
2,4-Dinitrophenylhydrazine, reagent grade, 97%
C6H6N4O4
A6003
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
D0700000
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
C22H29FO5
1176007
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
C22H29FO5
46165
Dexamethasone, VETRANAL, analytical standard
C22H29FO5
31375
Dexamethasone, tested according to Ph.Eur.
C22H29FO5
PHR1526
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
C22H29FO5
Y0001593
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
C22H29FO5
Y0001177
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
C22H29FO5
F0291 hBFGF, FGF-Basic, recombinant, expressed in E. coli, suitable for cell culture
44208
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
C6H14N2O2 · HCl
PHR1101
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C6H14N2O2 · HCl
67448
L-Lysine monohydrochloride, certified reference material, TraceCERT®
C6H14N2O2 · HCl
L0900000
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C6H14N2O2 · HCl
O4876
Oligomycin from Streptomyces diastatochromogenes, ≥90% total oligomycins basis (HPLC)
R8875
Rotenone, ≥95%
C23H22O6
45656
Rotenone, PESTANAL®, analytical standard
C23H22O6
P9767
Sodium palmitate, ≥98.5%
C16H31NaO2
PHR1187
Tretinoin, Pharmaceutical Secondary Standard; Certified Reference Material
C20H28O2
1674004
Tretinoin, United States Pharmacopeia (USP) Reference Standard
C20H28O2
T1850000
Tretinoin, European Pharmacopoeia (EP) Reference Standard
C20H28O2