EMAIL THIS PAGE TO A FRIEND
Nature communications
Large-scale analysis of lysine SUMOylation by SUMO remnant immunoaffinity profiling.
PMID 25391492
Abstract
Small ubiquitin-related modifiers (SUMO) are evolutionarily conserved ubiquitin-like proteins that regulate several cellular processes including cell cycle progression, intracellular trafficking, protein degradation and apoptosis. Despite the importance of protein SUMOylation in different biological pathways, the global identification of acceptor sites in complex cell extracts remains a challenge. Here we generate a monoclonal antibody that enriches for peptides containing SUMO remnant chains following tryptic digestion. We identify 954 SUMO3-modified lysine residues on 538 proteins and profile by quantitative proteomics the dynamic changes of protein SUMOylation following proteasome inhibition. More than 86% of these SUMOylation sites have not been reported previously, including 5 sites on the tumour suppressor parafibromin (CDC73). The modification of CDC73 at K136 affects its nuclear retention within PML nuclear bodies on proteasome inhibition. In contrast, a CDC73 K136R mutant translocates to the cytoplasm under the same conditions, further demonstrating the effectiveness of our method to characterize the dynamics of lysine SUMOylation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0267
2-Chloroacetamide, ≥98%
C2H4ClNO
22790
2-Chloroacetamide, ≥98.0% (HPLC)
C2H4ClNO
09830
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
CH5NO3
338818
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
H5NO
320145
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
H5NO
09859
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
H5NO
40205
Ammonium hydroxide solution, ≥25% NH3 in H2O, semiconductor grade PURANAL (Honeywell 17814)
H5NO
40291
Ammonium hydroxide solution, ≥25% NH3 in H2O, semiconductor grade VLSI PURANAL (Honeywell 17605)
H5NO
40321
Ammonium hydroxide solution, ≥25% NH3 in H2O, semiconductor grade SLSI PURANAL (Honeywell 17301)
H5NO
01-4362
Ammonium hydroxide solution, 28% NH3 in H2O, suitable for determination of toxic metals
H5NO
E3132
E-64, protease inhibitor
C15H27N5O5
49897
Formate standard for IC, 1.000 g/L in H2O, analytical standard
CH2O2
06473
Formic acid solution, BioUltra, 1.0 M in H2O
CH2O2
02739
Imidazole, ≥95.0% (HPLC), pharmaceutical impurity standard
C3H4N2
1336500
Imidazole, United States Pharmacopeia (USP) Reference Standard
C3H4N2
I0086000
Imidazole, European Pharmacopoeia (EP) Reference Standard
C3H4N2
PHR1180
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
C3H4N2
Y0001326
Ondansetron impurity E, European Pharmacopoeia (EP) Reference Standard
C3H4N2
243655
Sodium molybdate, ≥98%
MoNa2O4
737860
Sodium molybdate, anhydrous, powder, −100 mesh particle size, 99.9% trace metals basis
MoNa2O4
79299
Sodium tartrate dibasic solution, BioUltra, 1.5 M in H2O (colorless solution at 20 °C)
C4H4Na2O6
74564
Trifluoroacetic acid, analytical standard
C2HF3O2
U0600000
Urea, European Pharmacopoeia (EP) Reference Standard
CH4N2O
1706698
Urea, United States Pharmacopeia (USP) Reference Standard
CH4N2O
56180
Urea, analytical standard
CH4N2O
51465
Urea, tested according to Ph.Eur.
CH4N2O
08582
Urea solution, for microbiology, 40% in H2O
CH4N2O