EMAIL THIS PAGE TO A FRIEND
Cell death & disease
The Hippo pathway is controlled by Angiotensin II signaling and its reactivation induces apoptosis in podocytes.
PMID 25393475
Abstract
The Hippo pathway fulfills a crucial function in controlling the balance between proliferation, differentiation and apoptosis in cells. Recent studies showed that G protein-coupled receptors (GPCRs) serve as upstream regulators of Hippo signaling, that either activate or inactivate the Hippo pathway via the large tumor suppressor kinase (LATS) and its substrate, the co-transcription factor Yes-associated protein (YAP). In this study, we focused on the Angiotensin II type 1 receptor (AT1R), which belongs to the GPCR family and has an essential role in the control of blood pressure and water homeostasis. We found that Angiotensin II (Ang II) inactivates the pathway by decreasing the activity of LATS kinase; therefore, leading to an enhanced nuclear shuttling of unphosphorylated YAP in HEK293T cells. This shuttling of YAP is actin-dependent as disruption of the actin cytoskeleton inhibited dephosphorylation of LATS and YAP. Interestingly, in contrast to HEK293T cells, podocytes, which are a crucial component of the glomerular filtration barrier, display a predominant nuclear YAP localization in vivo and in vitro. Moreover, stimulation with Ang II did not alter Hippo pathway activity in podocytes, which show a deactivated pathway. Reactivation of the LATS kinase activity in podocytes resulted in an increased cytoplasmic YAP localization accompanied by a strong induction of apoptosis. Thus, our work indicates that the control of LATS activation and subsequent YAP localization is important for podocyte homeostasis and survival.