EMAIL THIS PAGE TO A FRIEND
Cardiovascular research
Absence of the calcium-binding protein, S100A1, confers pulmonary hypertension in mice associated with endothelial dysfunction and apoptosis.
PMID 25395393
Abstract
S100A1, a 10-kDa, Ca(2+)-binding protein, is expressed in endothelial cells (ECs) and binds eNOS. Its absence is associated with impaired production of nitric oxide (NO) and mild systemic hypertension. As endothelial dysfunction contributes to clinical and experimental pulmonary hypertension (PH), we investigated the impact of deleting S100A1 in mice, on pulmonary haemodynamics, endothelial function, NO production, associated signalling pathways, and apoptosis. Compared with wild-type (WT), S100A1-knock-out mice (KO) exhibited increased right ventricular (RV) weight/body weight ratio and elevated RV pressure in the absence of altered left ventricular filling pressures, accompanied by increase in wall thickness of muscularized pulmonary arteries and a reduction in microvascular perfusion. In isolated lung preparations, KO revealed reduced basal NO, blunted dose-responsiveness to acetylcholine, and augmented basal and angiotensin (AII)-induced pulmonary vascular resistance (R₀) compared with WT. Pre-treatment of KO lungs with S100A1 attenuated the AII-induced increase in pulmonary arterial pressure and R₀. S100A1-induced phosphorylation of eNOS, Akt, and ERK1/2 is attenuated in pulmonary EC of KO compared with WT. Basal and TNF-α-induced EC apoptosis is greater in KO vs. WT, and cell survival is enhanced by S100A1 treatment. Our data demonstrate that the absence of S100A1 results in PH by disruption of its normal capacity to (i) enhance pulmonary EC function by induction of eNOS activity and NO levels via Akt/ERK1/2 pathways and (ii) promote EC survival. The ability of exogenously administered S100A1 to rescue this phenotype makes it an attractive therapeutic target in the treatment of PH.