PloS one
Promoter methylation-mediated silencing of β-catenin enhances invasiveness of non-small cell lung cancer and predicts adverse prognosis.
PMID 25396757
Abstract
β-Catenin plays dual role in adhesion complex formation and the Wnt signaling pathway. Although β-catenin expression appears to be upregulated and Wnt signaling pathway is activated in the majority of cancers, its expression level seems to be lost in non-small cell lung cancer (NSCLC). We previously reported that the promoter of β-catenin was hypermethylated in two NSCLC cell lines. In the current study, we expanded our analysis for the methylation status of β-catenin promoter region and its protein expression in seven NSCLC cell lines and a series of 143 cases of primary human lung cancer with adjacent non-neoplastic tissues. Quantitative methylation specific PCR (qMSP) analysis showed methylation of β-catenin promoter region in five NSCLC cell lines, with increased β-catenin protein levels upon 5'-Aza-2'-deoxycytidine (5-aza-dC) treatment. The methylation status in SPC (methylated) and A549 (unmethylated) was confirmed by bisulfite sequencing PCR. 5-Aza-dC treatment inhibited invasiveness of SPC but not A549. Immunofluorescence analysis showed membranous β-catenin expression was lost in SPC and could be re-established by 5-aza-dC, while Wnt3a treatment led to nuclear translocation of β-catenin in both SPC and A549. Dual-luciferase assays indicated that 5-aza-dC treatment caused no significant increase in Wnt signaling activity compared with Wnt3a treatment. The effect of demethylation agent in SPC can be reversed by β-catenin depletion but not E-cadherin depletion which indicated that the methylation mediated β-catenin silencing might enhance NSCLC invasion and metastasis in an E-cadherin independent manner. Subsequent immunohistochemistry results further confirmed that β-catenin promoter hypermethylation correlated with loss of immunoreactive protein expression, positive lymph node metastasis, high TNM stage and poor prognosis. The present study implicates β-catenin promoter hypermethylation in the mechanism of epigenetic changes underlying NSCLC metastasis and progression, thus indicating the potential of β-catenin as a novel epigenetic target for the treatment of NSCLC patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S0252
Sulfobromophthalein disodium salt hydrate, used to study hepatocyte transport functions
C20H8Br4Na2O10S2 · xH2O