EMAIL THIS PAGE TO A FRIEND
Journal of cosmetic dermatology
Polyacrylamide gel 10xa0years experience: with particular reference to complications from filling of the body of the lip.
PMID 25399617
Abstract
A review of 242 facial treatments, in 86 different patients, with polyacrylamide gel (PAAG) was carried out by the author between 2003 and 2013. To evaluate by retrospective study the long-term outcomes of PAAG filling for facial contouring. To quantify adverse events and patient comfort in prior mixing 0.3xa0mL of 2% lidocaine/1xa0mL PAAG and decanting into a smaller volume syringe. Review of clinical records combined with a patient survey. 11 of 166 (6.6%) lip body infections; 1 of 202 (0.5%) in other sites. Addition of 2% lidocaine (55 treatments/28 patients) reduced lip body infections (8.7% to 5.7%, Pxa0