EMAIL THIS PAGE TO A FRIEND
Biomedical chromatography : BMC
Simultaneous quantification of sildenafil and N-desmethyl sildenafil in human plasma by UFLC coupled with ESI-MS/MS and pharmacokinetic and bioequivalence studies in Malay population.
PMID 25400284
Abstract
A simple, rapid, specific and reliable UFLC coupled with ESI-MSMS assay method to simultaneously quantify sildenafil and N-desmethyl sildenafil, with loperamide as internal standard, was developed. Chromatographic separation was performed on a Thermo Scientific Accucore C18 column with an isocratic mobile phase composed of 0.1% v/v formic acid in purified water-methanol (20:80, v/v), at a flow rate of 0.3 mL/min. Sildenafil, N-desmethyl sildenafil and loperamide were detected with proton adducts at m/z 475.4 > 58.2, 461.3 > 85.2 and 477.0 > 266.1 in multiple reaction monitoring positive mode, respectively. Both analytes and internal standard were extracted by diethyl ether. The method was validated over a linear concentration range of 10-800 ng/mL for sildenafil and 10-600 ng/mL for N-desmethyl sildenafil with correlation coefficient (r(2) ) ≥0.9976 for sildenafil and (r(2) ) ≥0.9992 for N-desmethyl sildenafil. The method was precise, accurate and stable. The proposed method was applied to study the bioequivalence between a 100 mg dose of two pharmaceutical products: Viagra (original) and Edyfil (generic) products. AUC0-t , Cmax and Tmax were 2285.79 ng h/mL, 726.10 ng/mL and 0.94 h for Viagra and 2363.25 ng h/mL, 713.91 ng/mL and 0.83 hour for Edyfil. The 90% confidence interval of these parameters of this study fall within the regulatory range of 80-125%, hence they are considered as bioequivalent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PZ0003
Sildenafil citrate salt, ≥98% (HPLC)
C22H30N6O4S ·C6H8O7