PloS one
Variable episomal silencing of a recombinant herpesvirus renders its encoded GFP an unreliable marker of infection in primary cells.
PMID 25402328
Abstract
The availability of reliable recombinant reporter virus systems has been a great boon to the study of Kaposi's sarcoma-associated herpesvirus (KSHV/HHV-8). Unexpectedly, we found that expression of the ostensibly constitutive green fluorescent protein (GFP) marker was progressively lost during unselected passage in primary rat mesenchymal precursor cells (MM), despite efficient maintenance of latent viral gene expression and episomal partitioning. This repression of EF1-α promoter-driven GFP expression appeared to be passage-dependent, however, since functionally immortalized MM cells derived from long serial passage retained stable expression of GFP following rKSHV.219 infection. Chromatin analysis of cultures that we had infected in parallel demonstrated an increase in repressive H3K27 tri-methylation across the viral episome with the exception of the LANA control region in MM cells infected at early rather than late passage post-isolation. The silencing of GFP expression in the MM cells was reversible in a dose-dependent fashion by the histone deacetylase inhibitor valproic acid, further implicating cellular silencing on incoming viral genomes, and underscoring potential differences in viral gene regulation between primary and functionally immortalized cells. Furthermore, using multispectral imaging flow cytometry, we also determined that the extent of GFP expression per cell among those that were positive did not correlate with the number of LANA dots per nucleus nor the extent of overall LANA expression per cell. This suggests a more complex mode of local gene regulation, rather than one that simply reflects the relative intracellular viral copy number. In sum, we have demonstrated the significant potential for false-negative data when using a constitutive marker gene as a sole means of evaluating herpesviral infection, especially in primary cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P6273
2-Propylpentanoic acid
C8H16O2
07-449
Anti-trimethyl-Histone H3 (Lys27) Antibody, Anti-trimethyl-Histone H3 (Lys27), also known as Anti-H3K27me3, is a highly published Rabbit Polyclonal Antibody. This protein A purified antibody is dot blot tested for trimethylated lysine 27 specificity and validated in WB, ChIP, ChIP-seq, ICC, IP.
04-745
Anti-trimethyl-Histone H3 (Lys4) Antibody, clone MC315, rabbit monoclonal, Anti-trimethyl-Histone H3 (Lys4) Antibody, clone MC315 is a rabbit monoclonal antibody for detection of trimethyl-Histone H3 (Lys4) also known as H3K4me3, Histone H3 (tri methyl K4) & has been validated in WB, ChIP, DB, Mplex, ChIP-seq.
33429
Doxycycline hyclate, VETRANAL, analytical standard
C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O
PHR1145
Doxycycline hyclate, Pharmaceutical Secondary Standard; Certified Reference Material
C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O
D3000000
Doxycycline hyclate, European Pharmacopoeia (EP) Reference Standard
C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O
1226003
Doxycycline hyclate, United States Pharmacopeia (USP) Reference Standard
C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O
PHR1061
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C8H16O2
1708707
Valproic acid, United States Pharmacopeia (USP) Reference Standard
C8H16O2
V0033000
Valproic acid, European Pharmacopoeia (EP) Reference Standard
C8H16O2
Y0001422
Valproic acid for system suitability, European Pharmacopoeia (EP) Reference Standard
C8H16O2