EMAIL THIS PAGE TO A FRIEND
Age (Dordrecht, Netherlands)
Multiple, but not traditional risk factors predict mortality in older people: the Concord Health and Ageing in Men Project.
PMID 25403157
Abstract
This study aims to identify the common risk factors for mortality in community-dwelling older men. A prospective population-based study was conducted with a median of 6.7xa0years of follow-up. Participants included 1705 men aged ≥70xa0years at baseline (2005-2007) living in the community in Sydney, Australia. Demographic information, lifestyle factors, health status, self-reported history of diseases, physical performance measures, blood pressure, height and weight, disability (activities of daily living (ADL) and instrumental ADLs, instrumental ADLs (IADLs)), cognitive status, depressive symptoms and blood analyte measures were considered. Cox regression analyses were conducted to model predictors delete time until of mortality. During follow-up, 461 men (27xa0%) died. Using Cox proportional hazards model, significant predictors of delete time to time to mortality included in the final model (p < 0.05) were older age, body mass index < 20xa0kgxa0m(2), high white cell count, anaemia, low albumin, current smoking, history of cancer, history of myocardial infarction, history of congestive heart failure, depressive symptoms and ADL and IADL disability and impaired chair stands. We found that overweight and obesity and/or being a lifelong non-drinker of alcohol were protective against mortality. Compared to men with less than or equal to one risk factor, the hazard ratio in men with three risk factors was 2.5; with four risk factors, it was 4.0; with five risk factors, it was 4.9; and for six or more risk factors, it was 11.4, respectively. We have identified common risk factors that predict mortality that may be useful in making clinical decisions among older people living in the community. Our findings suggest that, in primary care, screening and management of multiple risk factors are important to consider for extending survival, rather than simply considering individual risk factors in isolation. Some of the "traditional" risk factors for mortality in a younger population, including high blood pressure, hypercholesterolaemia, overweight and obesity and diabetes, were not independent predictors of mortality in this population of older men.