EMAIL THIS PAGE TO A FRIEND
Journal of cancer research and clinical oncology
Identification of FLOT2 as a novel target for microRNA-34a in melanoma.
PMID 25403318
Abstract
To confirm whether flotillin 2 (FLOT2) is a direct target of miR-34a and miR-34a/FLOT2 pathway plays a key role in melanoma proliferation and metastasis. First, miR-34a and FLOT2 expressions were both detected in human tissues and cell lines by qRT-PCR. Then, after transfection of mimics/inhibitor of miR-34a into melanoma cell lines, MTT, colony formation, scratch migration assays and transwell invasion assays were performed to evaluate the impact of miR-34a on cell proliferation and metastasis. Western blot, qRT-RCR and dual luciferase reporter gene assays were carried out to confirm whether FLOT2 is a direct target gene of miR-34a. In functional recovery experiments, proliferation and metastasis ability of WM35 and WM451 was tested after being co-transfected with miR-34a inhibitor/si-FLOT2 or miR-34a mimics/FLOT2 cDNA to confirm that FLOT2 is downregulated by miR-34a. The miR-34a significantly lower-expressed in metastasis melanoma tissues compared to in situ melanoma, nevi and normal skin whereas FLOT2 has an opposite trend. The level of miR-34a and FLOT2 in different melanoma cell lines was also tested and found that metastatic melanoma cell lines has lower miR-34a expression and higher FLOT2 expression compare to in situ melanoma cell line. MiR-34a overexpression profoundly inhibits WM451 cell proliferation and metastasis, whereas miR-34a reduction had a promoting effect to proliferation and metastasis of WM35. Results of Western blot, qRT-RCR and dual luciferase reporter gene assays revealed that FLOT2 is a direct target gene of miR-34a. Furthermore, overexpression/blockage of FLOT2 could attenuate effect of miR-34a overexpression/inhibition which indicated miR-34a suppresses melanoma biological behavior partially through FLOT2 inhibition. Our study confirmed that miR-34a is involved in the tumor inhibition of melanoma by directly targeting FLOT2 gene. This finding provides potential novel strategies for therapeutic interventions of melanoma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2501233
Anti-F2R antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405778
Anti-F2R antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2100740
Anti-F2R antibody produced in rabbit, affinity isolated antibody
HPA001396
Anti-FLOT2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300638
Anti-FLOT2 antibody produced in rabbit, affinity isolated antibody
SAB1405818
Anti-FLOT2, (N-terminal) antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
F1680
Anti-Flotillin 2 (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
F1805
Anti-Flotillin 2 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB2101635
Anti-NR1I2 (ab1) antibody produced in rabbit, affinity isolated antibody
AV39532
Anti-NR1I2 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV39738
Anti-NR1I2 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB1409956
Anti-NR1I2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1411574
Anti-NR1I2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
T5700 Anti-Thrombin Receptor antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
EHU008551 MISSION® esiRNA, esiRNA targeting human FLOT2 (esiRNA1)
EMU009561 MISSION® esiRNA, esiRNA targeting mouse Flot2 (esiRNA1)
SAB1402186
Monoclonal Anti-F2R antibody produced in mouse, clone 2C5, purified immunoglobulin, buffered aqueous solution
SAB1402776
Monoclonal Anti-FLOT2, (N-terminal) antibody produced in mouse, clone 3G6, purified immunoglobulin, buffered aqueous solution
SAB5300042
Monoclonal Anti-PAR1 antibody produced in mouse, clone 6A7H10, ascites fluid