EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Generic structures of cytotoxic liprotides: nano-sized complexes with oleic acid cores and shells of disordered proteins.
PMID 25403886
Abstract
The cytotoxic complex formed between α-lactalbumin and oleic acid (OA) has inspired many studies on protein-fatty acid complexes, but structural insight remains sparse. After having used small-angle X-ray scattering (SAXS) to obtain structural information, we present a new, generic structural model of cytotoxic protein-oleic acid complexes, which we have termed liprotides (lipids and partially denatured proteins). Twelve liprotides formed from seven structurally unrelated proteins and prepared by different procedures all displayed core-shell structures, each with a micellar OA core and a shell consisting of flexible, partially unfolded protein, which stabilizes the OA micelle. The common structure explains similar effects exerted on cells by different liprotides and is consistent with a cargo off-loading of the OA into cell membranes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O7501
Sodium oleate, ≥99%
C18H33NaO2
O3880
Sodium oleate, ≥95% (capillary GC)
C18H33NaO2