EMAIL THIS PAGE TO A FRIEND
International journal of oncology
Dual regulation of receptor tyrosine kinase genes EGFR and c-Met by the tumor-suppressive microRNA-23b/27b cluster in bladder cancer.
PMID 25405368
Abstract
Recent clinical trials of chemotherapeutics for advanced bladder cancer (BC) have shown limited benefits. Therefore, new prognostic markers and more effective treatment strategies are required. One approach to achieve these goals is through the analysis of RNA networks. Our recent studies of microRNA (miRNA) expression signatures revealed that the microRNA-23b/27b (miR-23b/27b) cluster is frequently downregulated in various types of human cancers. However, the functional role of the miR-23b/27b cluster in BC cells is still unknown. Thus, the aim of the present study was to investigate the functional significance of the miR-23b/27b cluster and its regulated molecular targets, with an emphasis on its contributions to BC oncogenesis and metastasis. The expression levels of the miR-23b/27b cluster were significantly reduced in BC clinical specimens. Restoration of mature miR-23b or miR-27b miRNAs significantly inhibited cancer cell migration and invasion, suggesting that these clustered miRNAs function as tumor suppressors. Gene expression data and in silico analysis demonstrated that the genes coding for the epidermal growth factor receptor (EGFR) and hepatocyte growth factor receptor (c-Met) were potential targets of the miR-23b/27b cluster. Luciferase reporter assays and western blotting demonstrated that EGFR and c-Met receptor trypsine kinases were directly regulated by these clustered miRNAs. We conclude that the decreased expression of the tumor-suppressive miR-23b/27b cluster enhanced cancer cell proliferation, migration and invasion in BC through direct regulation of EGFR and c-Met signaling pathways. Our data on RNA networks regulated by tumor-suppressive miR-23b/27b provide new insights into the potential mechanisms of BC oncogenesis and metastasis.