EMAIL THIS PAGE TO A FRIEND
Faraday discussions
Effect of double blocking layers at TiO2/Sb2S3 and Sb2S3/spiro-MeOTAD interfaces on photovoltaic performance.
PMID 25406977
Abstract
The effect of double blocking layers on the Sb(2)S(3)-sensitized all solid state solar cell are investigated. Thin layers of ZrO(2) (blocking layer 1, BL1) and ZnS (blocking layer 2, BL2) are introduced at the TiO(2)/Sb(2)S(3) and Sb(2)S(3)/hole transporting material (HTM) interfaces. The presence of BL1 is found to improve the open-circuit voltage (V(oc)) from 457 mV to 618 mV, whereas BL2 improves mainly short-circuit current density (J(sc)) from 11.1 mA cm(-2) to 14.0 mA cm(-2). Transient photovoltage confirms that the BL1 efficiently blocks charge recombination, responsible for the V(oc) enhancement, whereas the BL2 has little effect on suppression of charge recombination. Surface modification of Sb(2)S(3) by BL2, on the other hand, leads to recovery of Sb(2)S(3) from the surface oxidized Sb(2)O(5), which affects the J(sc) increment. The present results suggest that the TiO(2)/Sb(2)S(3) interface is the main pathway for charge recombination rather than the Sb(2)S(3)/HTM interface. Double blocking layers enhanced the power conversion efficiency by 30%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19422
1-Butanol, analytical standard
C4H10O
154679
1-Butanol, spectrophotometric grade, 99.5%
C4H10O
19420
1-Butanol, puriss. p.a., ACS reagent, ≥99.5% (GC)
C4H10O
442665
1-Butanol, analytical standard
C4H10O
PHR1209
1-Butanol, Pharmaceutical Secondary Standard; Certified Reference Material
C4H10O
38433
Diisopropoxytitanium bis(acetylacetonate) solution, purum, ~75% in isopropanol
C16H28O6Ti
S7026
Sodium thiosulfate
Na2O3S2
563188
Sodium thiosulfate, ≥99.99% trace metals basis
Na2O3S2
HT1005
Sodium thiosulfate solution, 2 g/dL in deionized water
Na2O3S2
77663
Terpineol, mixture of isomers, analytical standard
C10H18O
86480
Terpineol, mixture of isomers, anhydrous
C10H18O
T3407
Terpineol, ~95%
C10H18O
W304514
Terpineol, natural
C10H18O
325252
Titanium diisopropoxide bis(acetylacetonate), 75 wt. % in isopropanol
C16H28O6Ti
677469
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
O2Ti
718467
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
O2Ti
774510
Titanium(IV) oxide, nanowires, diam. × L ~100 nm × 10 μm
O2Ti
774529
Titanium(IV) oxide, nanowires, diam. × L ~10 nm × 10 μm
O2Ti
637254
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
O2Ti
232033
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
O2Ti
248576
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
O2Ti
634662
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
O2Ti
700355
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticle, <250 nm particle size (DLS), paste, 53-57 wt. % in diethylene glycol monobutyl ether/ethylene glycol, 99.9% trace metals basis
O2Ti
700347
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 33-37 wt. % in H2O, 99.5% trace metals basis
O2Ti
700339
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <100 nm particle size, dispersion, 48-52 wt. % in xylene, 99.9% trace metals basis
O2Ti
204730
Titanium(IV) oxide, rutile, 99.995% trace metals basis
O2Ti
204757
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
O2Ti
635057
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), 99.99% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
O2Ti
635049
Titanium(IV) oxide, rutile, <100>, (single crystal substrate), 99.99% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
O2Ti
635065
Titanium(IV) oxide, rutile, <110>, single crystal substrate, 99.99% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
O2Ti