EMAIL THIS PAGE TO A FRIEND
Radiation research
Age-dependent changes in DNA repair in radiation-exposed mice.
PMID 25409128
Abstract
Ionizing radiation (IR) is a well-known human carcinogen. Young and adult individuals are known to respond to radiation in a different manner. In this study, we analyzed changes in the spleen of juvenile (two-week-old), adult (two-month-old) and old (18-month-old) C57BL/6 male mice subjected to a whole-body exposure to 1 Gy of X rays. We measured the number of γ-H2AX foci and ATM protein levels as a reflection of the level of DNA double-strand breaks (DSBs), and found that old animals had a high frequency of occurrence of noninduced DSBs. Exposure to X rays resulted in a rapid increase in the number of DSBs in juvenile and adult animals at 6 h postirradiation followed by a return to preirradiated DSB values at 96 h postirradiation. No changes were observed in old animals. The analysis of the levels of proteins involved in DNA damage base excision and mismatch repair pathways, including KU70, RAD51, POL β, POL δ, POL ε, APE1 and MSH2 showed substantial age-dependent radiation-induced differences. Finally, we demonstrated that old animals had a higher background level of cell apoptosis compared to younger animals, but in contrast to younger animals, old animals were not able to commit spleen cells to apoptosis after being irradiated. Thus, spleen cells of old mice have a high level of spontaneous DNA damage, but they are not able to deal with additional radiation-induced damage as efficiently as younger animals, substantiating age-depending differences in radiation-induced DNA damage and repair response and its outcomes.
Related Materials