EMAIL THIS PAGE TO A FRIEND
Applied microbiology and biotechnology
Combined effects of nutrients and temperature on the production of fermentative aromas by Saccharomyces cerevisiae during wine fermentation.
PMID 25412578
Abstract
Volatile compounds produced by yeast during fermentation greatly influence the organoleptic qualities of wine. We developed a model to predict the combined effects of initial nitrogen and phytosterol content and fermentation temperature on the production of volatile compounds. We used a Box-Behnken design and response surface modeling to study the response of Lalvin EC1118® to these environmental conditions. Initial nitrogen content had the greatest influence on most compounds; however, there were differences in the value of fermentation parameters required for the maximal production of the various compounds. Fermentation parameters affected differently the production of isobutanol and isoamyl alcohol, although their synthesis involve the same enzymes and intermediate. We found differences in regulation of the synthesis of acetates of higher alcohols and ethyl esters, suggesting that fatty acid availability is the main factor influencing the synthesis of ethyl esters whereas the production of acetates depends on the activity of alcohol acetyltransferases. We also evaluated the effect of temperature on the total production of three esters by determining gas-liquid balances. Evaporation largely accounted for the effect of temperature on the accumulation of esters in liquid. Nonetheless, the metabolism of isoamyl acetate and ethyl octanoate was significantly affected by this parameter. We extended this study to other strains. Environmental parameters had a similar effect on aroma production in most strains. Nevertheless, the regulation of the synthesis of fermentative aromas was atypical in two strains: Lalvin K1M® and Affinity™ ECA5, which produces a high amount of aromatic compounds and was obtained by experimental evolution.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W269514
(±)-2-Methylbutyric acid, natural, ≥98%, FG
C5H10O2
08996
(−)-Ethyl L-lactate, analytical standard
C5H10O3
73117
1-Hexanol, analytical standard
C6H14O
52840
1-Hexanol, purum, ≥98.0% (GC)
C6H14O
471402
1-Hexanol, anhydrous, ≥99%
C6H14O
H13303
1-Hexanol, reagent grade, 98%
C6H14O
52828
1-Hexanol, ReagentPlus®, ≥99.5% (GC)
C6H14O
P13800
1-Phenylethanol, 98%
C8H10O
58448
2-Methyl-1-propanol, BioUltra, for molecular biology, ≥99.5% (GC)
C4H10O
82059
2-Methyl-1-propanol, analytical standard
C4H10O
58450
2-Methyl-1-propanol, puriss. p.a., ACS reagent, ≥99.5% (GC)
C4H10O
33064
2-Methyl-1-propanol, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
C4H10O
294829
2-Methyl-1-propanol, anhydrous, 99.5%
C4H10O
270466
2-Methyl-1-propanol, for HPLC, 99.5%
C4H10O
320048
2-Methyl-1-propanol, ACS reagent, ≥99.0%
C4H10O
538132
2-Methyl-1-propanol, 99.5%
C4H10O
30761
2-Methylbutyl acetate, analytical standard
C7H14O2
W364401
2-Methylbutyl acetate, 99%, FG
C7H14O2
W364428
2-Methylbutyl acetate, natural, ≥95%, FG
C7H14O2
W269506
2-Methylbutyric acid, ≥98%, FG
C5H10O2
193070
2-Methylbutyric acid, 98%
C5H10O2
77861
2-Phenylethanol, ≥99.0% (GC)
C8H10O
W268518
α-Methylbenzyl alcohol, ≥99%, FCC, FG
C8H10O
85451
β-Sitosterol, ≥70%
C29H50O
19215
Butyric acid, analytical standard
C4H8O2
B103500
Butyric acid, ≥99%
C4H8O2
W222100
Butyric acid, ≥99%, FG
C4H8O2
W222119
Butyric acid, natural, ≥99%, FCC, FG
C4H8O2
C0426000
Caprylic acid, European Pharmacopoeia (EP) Reference Standard
C8H16O2
PHR1202
Caprylic Acid (Octanoic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
C8H16O2