EMAIL THIS PAGE TO A FRIEND
BMC nephrology
Low serum calcium is associated with poor renal outcomes in chronic kidney disease stages 3-4 patients.
PMID 25412875
Abstract
Mineral disorders are associated with adverse renal outcomes in chronic kidney disease (CKD) patients. Previous studies have associated hypercalcemia and hypocalcemia with mortality; however, the association between serum calcium and renal outcome is not well-described. Whether adding calcium besides phosphorus or in the form of calcium-phosphorus (Ca×P) product into the model of survival analysis could improve the prediction of renal outcomes is not known. A prospective cohort of 2144 outpatients with CKD stages 3-4 was evaluated. Cox proportional hazard analysis was performed according to calcium quartiles. The mean calcium level was 9.2±0.7 mg/dL. Low serum calcium (<9.0 mg/dL) was associated with increased risk of requiring renal replacement therapy (RRT) (hazards ratio [HR]:2.12 (95% CI: 1.49-3.02, P<0.05) and rapid renal function progression (odds ratio [OR]: 1.65 (95% CI: 1.19-2.27, P<0.05) compared with high serum calcium (>9.8 mg/dL). Adding calcium into the survival model increased the integrated discrimination improvement by 0.80% (0.12%-1.91%) while calcium-phosphorus product did not improve risk prediction.The combination of high serum phosphorus (>4.2 mg/dL) and low serum calcium (<9.1 mg/dL) was associated with the highest risk of RRT (HR:2.31 (95% CI: 1.45-3.67, P<0.05). Low serum calcium is associated with increased risk of RRT and rapid renal function progression in CKD stage 3-4 patients. The integration of serum calcium and phosphorus, but not calcium-phosphorus product should be considered in a predictive model of renal outcome.