EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Gastrointestinal and liver physiology
Caspase-11 attenuates gastrointestinal inflammation and experimental colitis pathogenesis.
PMID 25414099
Abstract
Nucleotide-binding domain and leucine-rich repeat containing protein inflammasome formation plays an essential role in modulating immune system homeostasis in the gut. Recently, a caspase-11 noncanonical inflammasome has been characterized and appears to modulate many biological functions that were previously considered to be solely dependent on caspase-1 and the canonical inflammasome. To better elucidate the function of this noncanonical inflammasome during inflammatory bowel disease, experimental colitis was induced in wild-type and Casp11(-/-) mice utilizing dextran sulfate sodium (DSS). Here, we report that caspase-11 attenuates acute experimental colitis pathogenesis. Casp11(-/-) mice showed significantly increased morbidity and colon inflammation following DSS exposure. Subsequent cytokine analysis revealed that IL-1β and IL-18 levels in the colon were significantly reduced in the Casp11(-/-) mice compared with the wild-type animals. Additional mechanistic studies utilizing IL-1β and IL-18 reconstitution revealed that Casp11(-/-) hypersensitivity was associated with the loss of both of these cytokines. Bone marrow reconstitution experiments further revealed that caspase-11 gene expression and function in both hematopoietic- and nonhematopoietic-derived cells contribute to disease attenuation. Interestingly, unlike caspase-1, caspase-11 does not appear to influence relapsing remitting disease progression or the development of colitis-associated tumorigenesis. Together, these data identify caspase-11 as a critical factor protecting the host during acute DSS-induced colonic injury and inflammation but not during chronic inflammation and tumorigenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5486
Azoxymethane, 13.4 M, ≥98%
C2H6N2O