EMAIL THIS PAGE TO A FRIEND
British journal of anaesthesia
Effect of flumazenil on diaphragm electrical activation during weaning from mechanical ventilation after acute respiratory distress syndrome.
PMID 25416275
Abstract
Diaphragm electrical activation (EAdi) and the ratio of tidal volume to EAdi (VT/EAdi) may provide clinical information on neuroventilatory efficiency (NVE) in patients being weaned from mechanical ventilation. We tested the hypothesis that residual sedation could interfere with respiratory recovery, by assessing the effects of flumazenil on EAdi and VT/EAdi ratio. This observational study included 13 patients breathing with pressure-support ventilation (PSV) after a long period of controlled mechanical ventilation (i.e. >4 days) plus midazolam-based sedation for acute respiratory distress syndrome. EAdi and respiratory patterns were compared before and after a bolus of flumazenil, which was given because neurological status needed to be evaluated. Flumazenil induced a significant increase in EAdi [+71 (41-123)%, P=0.0002] and VT [+17 (8-32)%, P=0.0005], resulting in significantly decreased NVE [-34 (15-43)%]. The increased VT was significantly correlated with the increased EAdi (ρ=0.70, P=0.009). During weaning from mechanical ventilation, the diaphragmatic contribution to the breathing process may be reduced by residual midazolam-induced ventilatory depression. The increased EAdi with reversal of residual sedation was associated with a proportional increase in VT. These findings should be considered by the attending physician when interpreting daily EAdi and VT changes during weaning from mechanical ventilation.