EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Anionic glycolipids related to glucuronosyldiacylglycerol inhibit protein kinase Akt.
PMID 25417778
Abstract
New glucuronosyldiacylglycerol (GlcADG) analogues based on a 2-O-β-D-glucopyranosyl-sn-glycerol scaffold and carrying one or two acyl chains of different lengths have been synthesized as phosphatidylinositol 3-phosphate (PI3P) mimics targeting the protein kinase Akt. The Akt inhibitory effect of the prepared compounds was assayed using an in vitro kinase assay. The antiproliferative activity of the compounds was tested in the human ovarian carcinoma IGROV-1 cell line in which we found that two of them could inhibit proliferation, in keeping with the target inhibitory effect.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
37095
Staurosporine, VETRANAL, analytical standard
C28H26N4O3