EMAIL THIS PAGE TO A FRIEND
International dental journal
FDI policy statement on dental amalgam and the Minamata Convention on Mercury: adopted by the FDI General Assembly: 13 September 2014, New Delhi, India.
PMID 25417784